Learn More
2,4-Difluorophenylglyoxal hydrate, 95%, dry wt. basis, Thermo Scientific Chemicals
$204.41 - $831.81
Chemical Identifiers
CAS | 79784-36-4 |
---|---|
Molecular Formula | C8H4F2O2 |
Molecular Weight (g/mol) | 170.12 |
MDL Number | MFCD04038287 |
InChI Key | AZBQYELTMQNVST-UHFFFAOYSA-N |
Synonym | 2,4-difluorophenylglyoxal hydrate, 2-2,4-difluorophenyl-2-oxoacetaldehyde hydrate, 2,4-difluorophenylglyoxal monohydrate, benzeneacetaldehyde,2,4-difluoro-a-oxo, 2-2,4-difluorophenyl-2-oxoethanal, hydrate, 2-2,4-bis fluoranyl phenyl-2-oxidanylidene-ethanal hydrate |
PubChem CID | 2782300 |
IUPAC Name | 2-(2,4-difluorophenyl)-2-oxoacetaldehyde;hydrate |
SMILES | FC1=CC(F)=C(C=C1)C(=O)C=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL2026603
|
Thermo Scientific Chemicals
L2026603 |
1 g |
Each for $204.41
|
|
|||||
AAL2026606
|
Thermo Scientific Chemicals
L2026606 |
5 g |
Each for $831.81
|
|
|||||
Description
Used in the synthesis of biologically active 1,2,4-triazine derivatives. Also used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed in the synthesis of biologically active 1,2,4-triazine derivatives. Also used as pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemical Identifiers
79784-36-4 | |
170.12 | |
AZBQYELTMQNVST-UHFFFAOYSA-N | |
2782300 | |
FC1=CC(F)=C(C=C1)C(=O)C=O |
C8H4F2O2 | |
MFCD04038287 | |
2,4-difluorophenylglyoxal hydrate, 2-2,4-difluorophenyl-2-oxoacetaldehyde hydrate, 2,4-difluorophenylglyoxal monohydrate, benzeneacetaldehyde,2,4-difluoro-a-oxo, 2-2,4-difluorophenyl-2-oxoethanal, hydrate, 2-2,4-bis fluoranyl phenyl-2-oxidanylidene-ethanal hydrate | |
2-(2,4-difluorophenyl)-2-oxoacetaldehyde;hydrate |
Specifications
79784-36-4 | |
C8H4F2O2 | |
1 g | |
2,4-difluorophenylglyoxal hydrate, 2-2,4-difluorophenyl-2-oxoacetaldehyde hydrate, 2,4-difluorophenylglyoxal monohydrate, benzeneacetaldehyde,2,4-difluoro-a-oxo, 2-2,4-difluorophenyl-2-oxoethanal, hydrate, 2-2,4-bis fluoranyl phenyl-2-oxidanylidene-ethanal hydrate | |
AZBQYELTMQNVST-UHFFFAOYSA-N | |
2-(2,4-difluorophenyl)-2-oxoacetaldehyde;hydrate | |
2782300 | |
95% | |
(dry wt.) |
51°C to 54°C | |
MFCD04038287 | |
4387465 | |
Soluble in water. | |
FC1=CC(F)=C(C=C1)C(=O)C=O | |
170.12 | |
170.12 (Anhydrous) | |
2,4-Difluorophenylglyoxal hydrate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only