missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2',4'-Dihydroxyacetophenone, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 114805000

 View more versions of this product

Catalog No. AC114805000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

2', 4'-Dihydroxyacetophenone
89-84-9
100.0
Authentic
Plastic bottle
(HO)2C6H3COCH3
500g
1.18
2',4'-dihydroxyacetophenone, 2,4-dihydroxyacetophenone, 1-2,4-dihydroxyphenyl ethanone, resacetophenone, 4-acetylresorcinol, resoacetophenone, ethanone, 1-2,4-dihydroxyphenyl, 1-acetyl-2,4-dihydroxybenzene, resorcinol, 4-acetyl, 1-2,4-dihydroxyphenyl ethan-1-one
SULYEHHGGXARJS-UHFFFAOYSA-N
1-(2,4-dihydroxyphenyl)ethan-1-one
6990
152.15
98%
97.5
1.1800g/mL
97.5% min. (HPLC)
C8H8O3
MFCD00002279
08, 266
15, 8260
Solubility in water: 1g/L (20 c). Other solubilities: soluble in glacial acetic acid,soluble in most common organic solvents except,benzene,chloroform and ether
CC(=O)C1=CC=C(O)C=C1O
152.15
CHEBI:18414
98%

Chemical Identifiers

89-84-9
152.15
SULYEHHGGXARJS-UHFFFAOYSA-N
6990
1-(2,4-dihydroxyphenyl)ethan-1-one
C8H8O3
MFCD00002279
2',4'-dihydroxyacetophenone, 2,4-dihydroxyacetophenone, 1-2,4-dihydroxyphenyl ethanone, resacetophenone, 4-acetylresorcinol, resoacetophenone, ethanone, 1-2,4-dihydroxyphenyl, 1-acetyl-2,4-dihydroxybenzene, resorcinol, 4-acetyl, 1-2,4-dihydroxyphenyl ethan-1-one
CHEBI:18414
CC(=O)C1=CC=C(O)C=C1O
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 201-945-3

RTECSNumber : AM7525000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only