missing translation for 'onlineSavingsMsg'
Learn More

2',4'-Dihydroxypropiophenone, 99%

CAS: 5792-36-9 | C9H10O3 | 166.176 g/mol

$100.79 - $294.81

Chemical Identifiers

CAS 5792-36-9
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00002280
InChI Key LLBBBYLDTDJMNU-UHFFFAOYSA-N
Synonym 2',4'-dihydroxypropiophenone, 1-2,4-dihydroxyphenyl propan-1-one, 2,4-dihydroxypropiophenone, 4-propionylresorcinol, 1-propanone, 1-2,4-dihydroxyphenyl, respropiophenone, resorcinol fragment, 9, acmc-209m1c, 2',4'-dihydroxy-propiophenone, 2/',4/'-dihydroxypropiophenone
PubChem CID 79856
IUPAC Name 1-(2,4-dihydroxyphenyl)propan-1-one
SMILES CCC(=O)C1=C(C=C(C=C1)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2341414
View Documents
Thermo Scientific Chemicals
B2341414
25 g
Each for $100.79
Only null left
 
AAB2341422
View Documents
Thermo Scientific Chemicals
B2341422
100 g
Each for $294.81
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5792-36-9
166.176
LLBBBYLDTDJMNU-UHFFFAOYSA-N
79856
CCC(=O)C1=C(C=C(C=C1)O)O
C9H10O3
MFCD00002280
2',4'-dihydroxypropiophenone, 1-2,4-dihydroxyphenyl propan-1-one, 2,4-dihydroxypropiophenone, 4-propionylresorcinol, 1-propanone, 1-2,4-dihydroxyphenyl, respropiophenone, resorcinol fragment, 9, acmc-209m1c, 2',4'-dihydroxy-propiophenone, 2/',4/'-dihydroxypropiophenone
1-(2,4-dihydroxyphenyl)propan-1-one

Specifications

5792-36-9
C9H10O3
25 g
2',4'-dihydroxypropiophenone, 1-2,4-dihydroxyphenyl propan-1-one, 2,4-dihydroxypropiophenone, 4-propionylresorcinol, 1-propanone, 1-2,4-dihydroxyphenyl, respropiophenone, resorcinol fragment, 9, acmc-209m1c, 2',4'-dihydroxy-propiophenone, 2/',4/'-dihydroxypropiophenone
CCC(=O)C1=C(C=C(C=C1)O)O
166.176
166.18
2',4'-Dihydroxypropiophenone
98°C to 101°C
MFCD00002280
1238635
LLBBBYLDTDJMNU-UHFFFAOYSA-N
1-(2,4-dihydroxyphenyl)propan-1-one
79856
99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 227-329-4

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only