missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dimethoxybenzylamine, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 369950050

 View more versions of this product

Catalog No. AC369950050

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2, 4-Dimethoxybenzylamine
97.5
Colorless to Yellow
140°C
Authentic
Glass bottle
1.5480 to 1.55
MFCD00052393
1.113
QOWBXWFYRXSBAS-UHFFFAOYSA-N
(2,4-dimethoxyphenyl)methanamine
597250
98%
20781-20-8
100.0
1.1130g/mL
>110°C
97.5% min. (GC)
C9H13NO2
(CH3O)2C6H3CH2NH2
5 mL
2,4-dimethoxybenzylamine, 2,4-dimethoxyphenyl methanamine, 2,4-dimethoxy benzylamine, 1-2,4-dimethoxyphenyl methanamine, benzenemethanamine, 2,4-dimethoxy, 2,4-dimethoxybenzyl amine, 2,4-dimethoxyphenyl methylamine, 2, 4-dimethoxybenzylamine, pubchem7398, 2,4dimethoxybenzylamine
COC1=CC(=C(C=C1)CN)OC
167.21
167.21
Liquid

Chemical Identifiers

20781-20-8
167.21
QOWBXWFYRXSBAS-UHFFFAOYSA-N
597250
COC1=CC(=C(C=C1)CN)OC
C9H13NO2
MFCD00052393
2,4-dimethoxybenzylamine, 2,4-dimethoxyphenyl methanamine, 2,4-dimethoxy benzylamine, 1-2,4-dimethoxyphenyl methanamine, benzenemethanamine, 2,4-dimethoxy, 2,4-dimethoxybenzyl amine, 2,4-dimethoxyphenyl methylamine, 2, 4-dimethoxybenzylamine, pubchem7398, 2,4dimethoxybenzylamine
(2,4-dimethoxyphenyl)methanamine
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only