Learn More
2,4-Dimethoxypyrimidine-5-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals
$272.86 - $272.86
Chemical Identifiers
CAS | 936250-17-8 |
---|---|
Molecular Formula | C12H19BN2O4 |
Molecular Weight (g/mol) | 266.10 |
MDL Number | MFCD06795654 |
InChI Key | RKHZHTLJEUBARP-UHFFFAOYSA-N |
Synonym | 2,4-dimethoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2,4-dimethoxypyrimidine-5-boronic acid pinacol ester, 2,6-dimethoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2,4-dimethoxypyrimidine-5-boronic acid, pinacol ester, 2,4-dimethoxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrimidine, 2,4-dimethoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2,4-dimethoxypyrimidine-5-boronicacidpinacolester, 2,4-dimethoxyprimidine-5-boronic acid pinacol ester, 2,6-dimethoxypyrimidine-5-boronic acid, pinacol ester |
PubChem CID | 16218388 |
IUPAC Name | 2,4-dimethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine |
SMILES | COC1=NC=C(B2OC(C)(C)C(C)(C)O2)C(OC)=N1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC444930050
|
Thermo Scientific Chemicals
444930050 |
5 g |
Each for $272.86
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
936250-17-8 | |
266.10 | |
RKHZHTLJEUBARP-UHFFFAOYSA-N | |
16218388 | |
COC1=NC=C(B2OC(C)(C)C(C)(C)O2)C(OC)=N1 |
C12H19BN2O4 | |
MFCD06795654 | |
2,4-dimethoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2,4-dimethoxypyrimidine-5-boronic acid pinacol ester, 2,6-dimethoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2,4-dimethoxypyrimidine-5-boronic acid, pinacol ester, 2,4-dimethoxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrimidine, 2,4-dimethoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2,4-dimethoxypyrimidine-5-boronicacidpinacolester, 2,4-dimethoxyprimidine-5-boronic acid pinacol ester, 2,6-dimethoxypyrimidine-5-boronic acid, pinacol ester | |
2,4-dimethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine |
Specifications
936250-17-8 | |
96% min. (GC) | |
MFCD06795654 | |
2,4-dimethoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2,4-dimethoxypyrimidine-5-boronic acid pinacol ester, 2,6-dimethoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2,4-dimethoxypyrimidine-5-boronic acid, pinacol ester, 2,4-dimethoxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrimidine, 2,4-dimethoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidine, 2,4-dimethoxypyrimidine-5-boronicacidpinacolester, 2,4-dimethoxyprimidine-5-boronic acid pinacol ester, 2,6-dimethoxypyrimidine-5-boronic acid, pinacol ester | |
COC1=NC=C(B2OC(C)(C)C(C)(C)O2)C(OC)=N1 | |
266.10 | |
266.11 | |
2, 4-Dimethoxypyrimidine-5-boronic acid pinacol ester |
104°C to 109°C | |
C12H19BN2O4 | |
5 g | |
RKHZHTLJEUBARP-UHFFFAOYSA-N | |
2,4-dimethoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine | |
16218388 | |
97% |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
RUO – Research Use Only