missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethyl-1,3-thiazole-5-carboxylic acid, 97%, Thermo Scientific™
Chemical Identifiers
| CAS | 53137-27-2 |
|---|---|
| Molecular Formula | C6H7NO2S |
| Molecular Weight (g/mol) | 157.187 |
| InChI Key | MQGBARXPCXAFRZ-UHFFFAOYSA-N |
| Synonym | 2,4-dimethylthiazole-5-carboxylic acid, dimethyl-1,3-thiazole-5-carboxylic acid, 5-thiazolecarboxylic acid, 2,4-dimethyl, 2,4-dimethyl-5-thiazolecarboxylic acid, 2,4-dimethyl-thiazole-5-carboxylic acid, 2,4-dimethylthiazole-5-carboxylicacid, ksc269g8n, 2,4-dimethylthiazol-5-carboxylic acid, 2,4-dimethylthiazole-5carboxylic acid, 2 4-dimethylthiazole-5-carboxylic acid |
| PubChem CID | 736488 |
| IUPAC Name | 2,4-dimethyl-1,3-thiazole-5-carboxylic acid |
| SMILES | CC1=C(SC(=N1)C)C(=O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
CC05801DA
|
Thermo Scientific Chemicals
CC05801DA |
1g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
CC05801EA
|
Thermo Scientific Chemicals
CC05801EA |
10g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 53137-27-2 | |
| 157.187 | |
| 2,4-dimethylthiazole-5-carboxylic acid, dimethyl-1,3-thiazole-5-carboxylic acid, 5-thiazolecarboxylic acid, 2,4-dimethyl, 2,4-dimethyl-5-thiazolecarboxylic acid, 2,4-dimethyl-thiazole-5-carboxylic acid, 2,4-dimethylthiazole-5-carboxylicacid, ksc269g8n, 2,4-dimethylthiazol-5-carboxylic acid, 2,4-dimethylthiazole-5carboxylic acid, 2 4-dimethylthiazole-5-carboxylic acid | |
| 2,4-dimethyl-1,3-thiazole-5-carboxylic acid |
| C6H7NO2S | |
| MQGBARXPCXAFRZ-UHFFFAOYSA-N | |
| 736488 | |
| CC1=C(SC(=N1)C)C(=O)O |
Specifications
| 53137-27-2 | |
| Amber Glass Bottle | |
| 1g | |
| MQGBARXPCXAFRZ-UHFFFAOYSA-N | |
| 2,4-dimethyl-1,3-thiazole-5-carboxylic acid | |
| 736488 | |
| 97% |
| 97% | |
| C6H7NO2S | |
| 2,4-dimethylthiazole-5-carboxylic acid, dimethyl-1,3-thiazole-5-carboxylic acid, 5-thiazolecarboxylic acid, 2,4-dimethyl, 2,4-dimethyl-5-thiazolecarboxylic acid, 2,4-dimethyl-thiazole-5-carboxylic acid, 2,4-dimethylthiazole-5-carboxylicacid, ksc269g8n, 2,4-dimethylthiazol-5-carboxylic acid, 2,4-dimethylthiazole-5carboxylic acid, 2 4-dimethylthiazole-5-carboxylic acid | |
| CC1=C(SC(=N1)C)C(=O)O | |
| 157.187 | |
| 157.19 | |
| 2, 4-Dimethyl-1, 3-thiazole-5-carboxylic acid |