missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4-Dimethyl-1-heptene 98.0+%, TCI America™
$97.89 - $268.76
Chemical Identifiers
CAS | 19549-87-2 |
---|---|
Molecular Formula | C9H18 |
Molecular Weight (g/mol) | 126.24 |
MDL Number | MFCD00059250 |
InChI Key | CZGAOHSMVSIJJZ-UHFFFAOYNA-N |
PubChem CID | 123385 |
IUPAC Name | 2,4-dimethylhept-1-ene |
SMILES | CCCC(C)CC(C)=C |
Chemical Identifiers
19549-87-2 | |
126.24 | |
CZGAOHSMVSIJJZ-UHFFFAOYNA-N | |
2,4-dimethylhept-1-ene |
C9H18 | |
MFCD00059250 | |
123385 | |
CCCC(C)CC(C)=C |
Specifications
19549-87-2 | |
134°C | |
MFCD00059250 | |
3295 | |
CCCC(C)CC(C)=C | |
126.24 | |
126.24 | |
Liquid |
Colorless | |
C9H18 | |
1 mL | |
CZGAOHSMVSIJJZ-UHFFFAOYNA-N | |
2,4-dimethylhept-1-ene | |
123385 | |
≥98.0% (GC) | |
2,4-Dimethyl-1-heptene |
Safety and Handling
EINECSNumber : (2)-0025
TSCA : No