missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethyl-2,4-pentanediol 96.0+%, TCI America™
$654.29 - $2289.06
Chemical Identifiers
| CAS | 24892-49-7 |
|---|---|
| Molecular Formula | C7H16O2 |
| Molecular Weight (g/mol) | 132.20 |
| MDL Number | MFCD00012306 |
| InChI Key | DBTGFWMBFZBBEF-UHFFFAOYSA-N |
| PubChem CID | 141153 |
| IUPAC Name | 2,4-dimethylpentane-2,4-diol |
| SMILES | CC(C)(O)CC(C)(C)O |
Chemical Identifiers
| 24892-49-7 | |
| 132.20 | |
| DBTGFWMBFZBBEF-UHFFFAOYSA-N | |
| 2,4-dimethylpentane-2,4-diol |
| C7H16O2 | |
| MFCD00012306 | |
| 141153 | |
| CC(C)(O)CC(C)(C)O |
Specifications
| 24892-49-7 | |
| 100°C | |
| MFCD00012306 | |
| DBTGFWMBFZBBEF-UHFFFAOYSA-N | |
| 2,4-dimethylpentane-2,4-diol | |
| 141153 | |
| ≥96.0% (GC) | |
| 2,4-Dimethyl-2,4-pentanediol |
| Yellow | |
| C7H16O2 | |
| 5 mL | |
| CC(C)(O)CC(C)(C)O | |
| 132.20 | |
| 132.20 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0240
TSCA : No