missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,4-Dimethyl-3-pentanol 99.0+%, TCI America™
$57.79 - $473.48
Chemical Identifiers
| CAS | 600-36-2 |
|---|---|
| Molecular Formula | C7H16O |
| Molecular Weight (g/mol) | 116.204 |
| MDL Number | MFCD00008921 |
| InChI Key | BAYAKMPRFGNNFW-UHFFFAOYSA-N |
| Synonym | 2,4-dimethyl-3-pentanol, diisopropylcarbinol, diisopropylmethanol, 3-pentanol, 2,4-dimethyl, di-iso-propylcarbinol, 2,4-dimethyl-3-hydroxypentane, unii-n54k81p01f, g00025-watson-int, ethyl tert-butyl carbinol, acmc-1adj1 |
| PubChem CID | 11752 |
| IUPAC Name | 2,4-dimethylpentan-3-ol |
| SMILES | CC(C)C(C(C)C)O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D238725ML
|
TCI America
D238725ML |
25 mL |
Each for $57.79
|
|
|||||
|
D2387500ML
|
TCI America
D2387500ML |
500 mL |
Each for $473.48
|
|
|||||
Chemical Identifiers
| 600-36-2 | |
| 116.204 | |
| BAYAKMPRFGNNFW-UHFFFAOYSA-N | |
| 11752 | |
| CC(C)C(C(C)C)O |
| C7H16O | |
| MFCD00008921 | |
| 2,4-dimethyl-3-pentanol, diisopropylcarbinol, diisopropylmethanol, 3-pentanol, 2,4-dimethyl, di-iso-propylcarbinol, 2,4-dimethyl-3-hydroxypentane, unii-n54k81p01f, g00025-watson-int, ethyl tert-butyl carbinol, acmc-1adj1 | |
| 2,4-dimethylpentan-3-ol |
Specifications
| 600-36-2 | |
| 136°C | |
| MFCD00008921 | |
| 1987 | |
| BAYAKMPRFGNNFW-UHFFFAOYSA-N | |
| 2,4-dimethylpentan-3-ol | |
| 11752 | |
| ≥99.0% (GC) | |
| 2,4-Dimethyl-3-pentanol |
| Colorless | |
| C7H16O | |
| 25 mL | |
| 2,4-dimethyl-3-pentanol, diisopropylcarbinol, diisopropylmethanol, 3-pentanol, 2,4-dimethyl, di-iso-propylcarbinol, 2,4-dimethyl-3-hydroxypentane, unii-n54k81p01f, g00025-watson-int, ethyl tert-butyl carbinol, acmc-1adj1 | |
| CC(C)C(C(C)C)O | |
| 116.204 | |
| 116.20 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0217
TSCA : Yes