missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dimethylbenzoic acid, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1058918

 View more versions of this product

Catalog No. AAA1058918


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2,4-Dimethylbenzoic acid
611-01-8
50 g
C9H10O2
971589
BKYWPNROPGQIFZ-UHFFFAOYSA-N
2,4-dimethylbenzoic acid
11897
150.18
124°C to 126°C
267°C (727 mmHg)
Odorless
MFCD00002480
benzoic acid, 2,4-dimethyl, 4-carboxy-1,3-dimethylbenzene, 2,4-dimethyl benzoic acid, m-xylene-4-carboxylic acid, 2,4-dimethyl-benzoic acid, 2,4-dimethylbenzoicacid, m-xylylic acid, pubchem15441, 2,4 dimethylbenzoic acid, benzoic acid,4-dimethyl
CC1=CC=C(C(O)=O)C(C)=C1
150.18
CHEBI:64811
98%

Chemical Identifiers

611-01-8
150.18
BKYWPNROPGQIFZ-UHFFFAOYSA-N
11897
2,4-dimethylbenzoic acid
C9H10O2
MFCD00002480
benzoic acid, 2,4-dimethyl, 4-carboxy-1,3-dimethylbenzene, 2,4-dimethyl benzoic acid, m-xylene-4-carboxylic acid, 2,4-dimethyl-benzoic acid, 2,4-dimethylbenzoicacid, m-xylylic acid, pubchem15441, 2,4 dimethylbenzoic acid, benzoic acid,4-dimethyl
CHEBI:64811
CC1=CC=C(C(O)=O)C(C)=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

EINECSNumber : 210-246-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only