missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dimethylbenzoic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 160320100

 View more versions of this product

Catalog No. AC160320100


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2, 4-Dimethylbenzoic acid
611-01-8
97.5
10 g
Authentic
Crystalline Powder
C9H10O2
MFCD00002480
benzoic acid, 2,4-dimethyl, 4-carboxy-1,3-dimethylbenzene, 2,4-dimethyl benzoic acid, m-xylene-4-carboxylic acid, 2,4-dimethyl-benzoic acid, 2,4-dimethylbenzoicacid, m-xylylic acid, pubchem15441, 2,4 dimethylbenzoic acid, benzoic acid,4-dimethyl
CC1=CC=C(C(O)=O)C(C)=C1
150.18
CHEBI:64811
98%
123°C to 127°C
White
100.0
267°C (727.0 mmHg)
Glass bottle
98%
(CH3)2C6H3CO2H
09, 531
BKYWPNROPGQIFZ-UHFFFAOYSA-N
2,4-dimethylbenzoic acid
11897
150.18

Chemical Identifiers

611-01-8
150.18
BKYWPNROPGQIFZ-UHFFFAOYSA-N
11897
2,4-dimethylbenzoic acid
C9H10O2
MFCD00002480
benzoic acid, 2,4-dimethyl, 4-carboxy-1,3-dimethylbenzene, 2,4-dimethyl benzoic acid, m-xylene-4-carboxylic acid, 2,4-dimethyl-benzoic acid, 2,4-dimethylbenzoicacid, m-xylylic acid, pubchem15441, 2,4 dimethylbenzoic acid, benzoic acid,4-dimethyl
CHEBI:64811
CC1=CC=C(C(O)=O)C(C)=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-246-2

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only