Learn More
2,4-Dimethylpentane, 98+%
CAS: 108-08-7 | C7H16 | 100.205 g/mol
$428.97 - $428.97
Chemical Identifiers
| CAS | 108-08-7 |
|---|---|
| Molecular Formula | C7H16 |
| Molecular Weight (g/mol) | 100.205 |
| MDL Number | MFCD00008945 |
| InChI Key | BZHMBWZPUJHVEE-UHFFFAOYSA-N |
| PubChem CID | 7907 |
| IUPAC Name | 2,4-dimethylpentane |
| SMILES | CC(C)CC(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL0357009
|
Thermo Scientific Chemicals
L0357009 |
10 g |
Each for $428.97
|
|
|||||
Description
It is not only used for synthesizing WeiNiGuan, but also used in binder and coating industry. It is used for synthesizing resin fiber. It is also used as intermediates and adhesives in oil drop coagulation thickener.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is not only used for synthesizing WeiNiGuan, but also used in binder and coating industry. It is used for synthesizing resin fiber. It is also used as intermediates and adhesives in oil drop coagulation thickener.
Solubility
Soluble in acetone, benzene, chloroform, alcohol and ether.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Chemical Identifiers
| 108-08-7 | |
| 100.205 | |
| BZHMBWZPUJHVEE-UHFFFAOYSA-N | |
| 2,4-dimethylpentane |
| C7H16 | |
| MFCD00008945 | |
| 7907 | |
| CC(C)CC(C)C |
Specifications
| 108-08-7 | |
| 0.67 | |
| −12°C (10°F) | |
| 1.382 | |
| 10 g | |
| 1696855 | |
| Soluble in acetone,benzene,chloroform,alcohol and ether. | |
| CC(C)CC(C)C | |
| 100.205 | |
| 100.21 | |
| 2,4-Dimethylpentane |
| ∼-123°C | |
| 79°C to 80°C | |
| C7H16 | |
| MFCD00008945 | |
| UN1206 | |
| pentane, 2,4-dimethyl, unii-4jt8q9qohi, diisopropylmethane, 4jt8q9qohi, 2,4-dimethylpentane, standard for gc, dimethylpentane, heptan und isomere, 2,4-dimethyl-pentan, pentane,2,4-dimethyl, 2,4-dimethylpentane | |
| BZHMBWZPUJHVEE-UHFFFAOYSA-N | |
| 2,4-dimethylpentane | |
| 7907 | |
| ≥98% |
Safety and Handling
GHS H Statement
H225-H304-H315-H336
Highly flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes skin irritation.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P312-P331-P332+P313-P363-P370+P378q-P501c
H225-H304-H315-H319-H336
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HEPTANES
EINECSNumber : 203-548-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only