missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,4-Dimethylphenylmagnesium bromide, 0.5M in 2-MeTHF, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H54134AE

Catalog No. AAH54134AE


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Reacts with water.

Notes
Air and moisture sensitive. Store
Specifications

Specifications

2,4-Dimethylphenylmagnesium bromide
C8H9BrMg
100 mL
Air and moisture sensitive; May form a precipitate
Reacts with water.
CC1=CC=C([Mg]Br)C(C)=C1
209.37
209.37
34589-46-3
MFCD01311493
UN2924
2,4-dimethylphenylmagnesium bromide, 2,4-dimethylphenylmagnesium bromide solution, hpgwkalajlxsdh-uhfffaoysa-m, 2,4-dimethylphenylmagne-sium bromide, 2,4-dimethylphenyl magnesium bromide, magnesium,1,3-dimethylbenzene-6-ide,bromide, 2,4-dimethylphenylmagnesium bromide, 0.5m in 2-methf, 2,4-dimethylphenylmagnesium bromide 0.5 m in tetrahydrofuran, 2,4-dimethylphenylmagnesium bromide solution, 0.5 m in thf, grignard reagent
HPGWKALAJLXSDH-UHFFFAOYSA-M
magnesium;1,3-dimethylbenzene-6-ide;bromide
13637491
0.5M in 2-MeTHF

Chemical Identifiers

34589-46-3
209.37
HPGWKALAJLXSDH-UHFFFAOYSA-M
13637491
CC1=CC=C([Mg]Br)C(C)=C1
C8H9BrMg
MFCD01311493
2,4-dimethylphenylmagnesium bromide, 2,4-dimethylphenylmagnesium bromide solution, hpgwkalajlxsdh-uhfffaoysa-m, 2,4-dimethylphenylmagne-sium bromide, 2,4-dimethylphenyl magnesium bromide, magnesium,1,3-dimethylbenzene-6-ide,bromide, 2,4-dimethylphenylmagnesium bromide, 0.5m in 2-methf, 2,4-dimethylphenylmagnesium bromide 0.5 m in tetrahydrofuran, 2,4-dimethylphenylmagnesium bromide solution, 0.5 m in thf, grignard reagent
magnesium;1,3-dimethylbenzene-6-ide;bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302-H314-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

TSCA : No

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions