Learn More
2,4-Dimethylthiazole-5-sulfonyl chloride, 97%
CAS: 80466-80-4 | C5H6ClNO2S2 | 211.678 g/mol
$68.14 - $670.82
Chemical Identifiers
| CAS | 80466-80-4 |
|---|---|
| Molecular Formula | C5H6ClNO2S2 |
| Molecular Weight (g/mol) | 211.678 |
| MDL Number | MFCD00052746 |
| InChI Key | GFFJSTHQILQFNQ-UHFFFAOYSA-N |
| Synonym | 2,4-dimethyl-1,3-thiazole-5-sulfonylchloride, 2,4-dimethylthiazole-5-sulfonyl chloride, 2,4-dimethyl-thiazole-5-sulfonyl chloride, dimethyl-1,3-thiazole-5-sulfonyl chloride, 2,4-dimethyl-5-thiazolesulfonyl chloride, 5-thiazolesulfonyl chloride, 2,4-dimethyl, 2,4-dimethyl-1,3-thiazole-5-sulphonyl chloride, 5-thiazolesulfonylchloride, 2,4-dimethyl, 2,4-dimethylthiazol-5-ylsulphonyl chloride |
| PubChem CID | 2776248 |
| IUPAC Name | 2,4-dimethyl-1,3-thiazole-5-sulfonyl chloride |
| SMILES | CC1=C(SC(=N1)C)S(=O)(=O)Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH54578MD
|
Thermo Scientific Chemicals
H54578MD |
250 mg |
Each for $68.14
|
|
|||||
|
AAH5457803
|
Thermo Scientific Chemicals
H5457803 |
1 g |
Each for $190.66
|
|
|||||
|
AAH5457806
|
Thermo Scientific Chemicals
H5457806 |
5 g |
Each for $670.82
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 80466-80-4 | |
| 211.678 | |
| GFFJSTHQILQFNQ-UHFFFAOYSA-N | |
| 2776248 | |
| CC1=C(SC(=N1)C)S(=O)(=O)Cl |
| C5H6ClNO2S2 | |
| MFCD00052746 | |
| 2,4-dimethyl-1,3-thiazole-5-sulfonylchloride, 2,4-dimethylthiazole-5-sulfonyl chloride, 2,4-dimethyl-thiazole-5-sulfonyl chloride, dimethyl-1,3-thiazole-5-sulfonyl chloride, 2,4-dimethyl-5-thiazolesulfonyl chloride, 5-thiazolesulfonyl chloride, 2,4-dimethyl, 2,4-dimethyl-1,3-thiazole-5-sulphonyl chloride, 5-thiazolesulfonylchloride, 2,4-dimethyl, 2,4-dimethylthiazol-5-ylsulphonyl chloride | |
| 2,4-dimethyl-1,3-thiazole-5-sulfonyl chloride |
Specifications
| 80466-80-4 | |
| C5H6ClNO2S2 | |
| 250 mg | |
| Moisture sensitive | |
| GFFJSTHQILQFNQ-UHFFFAOYSA-N | |
| 2,4-dimethyl-1,3-thiazole-5-sulfonyl chloride | |
| 2776248 | |
| 97% |
| 82°C to 84°C (0.3 mmHg) | |
| MFCD00052746 | |
| UN3265 | |
| 2,4-dimethyl-1,3-thiazole-5-sulfonylchloride, 2,4-dimethylthiazole-5-sulfonyl chloride, 2,4-dimethyl-thiazole-5-sulfonyl chloride, dimethyl-1,3-thiazole-5-sulfonyl chloride, 2,4-dimethyl-5-thiazolesulfonyl chloride, 5-thiazolesulfonyl chloride, 2,4-dimethyl, 2,4-dimethyl-1,3-thiazole-5-sulphonyl chloride, 5-thiazolesulfonylchloride, 2,4-dimethyl, 2,4-dimethylthiazol-5-ylsulphonyl chloride | |
| CC1=C(SC(=N1)C)S(=O)(=O)Cl | |
| 211.678 | |
| 211.68 | |
| 2,4-Dimethylthiazole-5-sulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only