Learn More
2,4-Dinitrobenzenesulfonic acid sodium salt, 97%, Thermo Scientific Chemicals
$52.78 - $152.11
Chemical Identifiers
CAS | 885-62-1 |
---|---|
Molecular Formula | C6H3N2NaO7S |
Molecular Weight (g/mol) | 270.15 |
MDL Number | MFCD00007471,MFCD10567393 |
InChI Key | GSBYVRKLPCSLNV-UHFFFAOYSA-M |
Synonym | sodium 2,4-dinitrobenzenesulfonate hydrate, acmc-209quh, sodium dnbs hydrate, 2,4-dinitrobenzenesulfonic acid sodium salt hydrate |
PubChem CID | 70700115 |
IUPAC Name | sodium;2,4-dinitrobenzenesulfonate;hydrate |
SMILES | [Na+].[O-][N+](=O)C1=CC=C(C(=C1)[N+]([O-])=O)S([O-])(=O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1849306
|
Thermo Scientific Chemicals
A1849306 |
5 g |
Each for $52.78
|
|
|||||
AAA1849314
|
Thermo Scientific Chemicals
A1849314 |
25 g |
Each for $152.11
|
|
|||||
Description
2,4-Dinitrobenzenesulfonic acid sodium salt is used in the synthesis of aromatic dimethylene spacer-inserted cyclodextrin derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,4-Dinitrobenzenesulfonic acid sodium salt is used in the synthesis of aromatic dimethylene spacer-inserted cyclodextrin derivatives.
Solubility
Soluble in water and methanol.
Notes
Hygroscopic. Moisture sensitive. Incompatible with strong oxidizing agents and strong bases.
Chemical Identifiers
885-62-1 | |
270.15 | |
GSBYVRKLPCSLNV-UHFFFAOYSA-M | |
70700115 | |
[Na+].[O-][N+](=O)C1=CC=C(C(=C1)[N+]([O-])=O)S([O-])(=O)=O |
C6H3N2NaO7S | |
MFCD00007471,MFCD10567393 | |
sodium 2,4-dinitrobenzenesulfonate hydrate, acmc-209quh, sodium dnbs hydrate, 2,4-dinitrobenzenesulfonic acid sodium salt hydrate | |
sodium;2,4-dinitrobenzenesulfonate;hydrate |
Specifications
885-62-1 | |
MFCD00007471,MFCD10567393 | |
6676807 | |
Soluble in water and methanol. | |
[Na+].[O-][N+](=O)C1=CC=C(C(=C1)[N+]([O-])=O)S([O-])(=O)=O | |
270.15 | |
270.15 | |
2,4-Dinitrobenzenesulfonic acid sodium salt |
C6H3N2NaO7S | |
5 g | |
sodium 2,4-dinitrobenzenesulfonate hydrate, acmc-209quh, sodium dnbs hydrate, 2,4-dinitrobenzenesulfonic acid sodium salt hydrate | |
GSBYVRKLPCSLNV-UHFFFAOYSA-M | |
sodium;2,4-dinitrobenzenesulfonate;hydrate | |
70700115 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 212-943-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only