missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,4-Dinitrophenylhydrazine, Spectrum™ Chemical
DI149, 119-26-6, C6H6N4O4
Supplier: Spectrum Chemical Mfg Cor DI149500GM
Description
Spectrum™ Chemical 2,4-Dinitrophenylhydrazine is a chemical compound that is usually supplied as a wet red-to-orange solid that is shock sensitive. It is used in Brady′s test to detect the carbonyl functionality of an aldehyde or ketone functional group.Specifications
119-26-6 | |
Fiber Drum | |
500 g | |
HORQAOAYAYGIBM-UHFFFAOYSA-N | |
(2,4-dinitrophenyl)hydrazine | |
65 to 70% | |
Crystalline Powder or Lumps |
100% | |
C6H6N4O4 | |
UN1325 | |
NNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O | |
198.14 | |
Reagent |
Chemical Identifiers
119-26-6 | |
198.14 | |
(2,4-dinitrophenyl)hydrazine |
C6H6N4O4 | |
HORQAOAYAYGIBM-UHFFFAOYSA-N | |
NNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O |