missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[(4-Fluorophenyl)thio]acetic acid, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals KM09206DA

 View more versions of this product

Catalog No. KM09206DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

2-[(4-Fluorophenyl)thio]acetic acid
97.0
Amber Glass Bottle
FC6H4SCH2CO2H
1g
HQEROVXUKINLPI-UHFFFAOYSA-N
2-(4-fluorophenyl)sulfanylacetic acid
736197
97%
332-51-4
97%
C8H7FO2S
MFCD00219825
4-fluorophenylthio acetic acid, 2-4-fluorophenylthio acetic acid, 2-4-fluorophenyl thio acetic acid, 4-fluorophenyl thio acetic acid, 4-fluorophenyl sulfanyl acetic acid, maybridge1_004918, acmc-20apn5, 4-fluoro-phenylsulfanyl-acetic acid, 2-4-fluorophenyl sulfanylacetic acid
C1=CC(=CC=C1F)SCC(=O)O
186.2
186.21

Chemical Identifiers

332-51-4
186.2
HQEROVXUKINLPI-UHFFFAOYSA-N
736197
C1=CC(=CC=C1F)SCC(=O)O
C8H7FO2S
MFCD00219825
4-fluorophenylthio acetic acid, 2-4-fluorophenylthio acetic acid, 2-4-fluorophenyl thio acetic acid, 4-fluorophenyl thio acetic acid, 4-fluorophenyl sulfanyl acetic acid, maybridge1_004918, acmc-20apn5, 4-fluoro-phenylsulfanyl-acetic acid, 2-4-fluorophenyl sulfanylacetic acid
2-(4-fluorophenyl)sulfanylacetic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions