Learn More
2,4-Hexadienal, predominantly trans,trans, 95%, Thermo Scientific Chemicals
$49.14 - $319.58
Chemical Identifiers
CAS | 142-83-6 |
---|---|
Molecular Formula | C6H8O |
Molecular Weight (g/mol) | 96.129 |
MDL Number | MFCD00007004 |
InChI Key | BATOPAZDIZEVQF-MQQKCMAXSA-N |
Synonym | 2,4-hexadienal, hexa-2,4-dienal, sorbaldehyde, sorbic aldehyde, trans,trans-2,4-hexadienal, 2e,4e-hexa-2,4-dienal, 2,4-hexadienal, 2e,4e, 2,4-hexadienal, e,e, 2-propyleneacrolein, 3-propyleneacrolein |
PubChem CID | 637564 |
ChEBI | CHEBI:82334 |
IUPAC Name | (2E,4E)-hexa-2,4-dienal |
SMILES | CC=CC=CC=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1320006
|
Thermo Scientific Chemicals
A1320006 |
5 g |
Each for $49.14
|
|
|||||
AAA1320014
|
Thermo Scientific Chemicals
A1320014 |
25 g |
Each for $119.62
|
|
|||||
AAA1320022
|
Thermo Scientific Chemicals
A1320022 |
100 g |
Each for $319.58
|
|
|||||
Description
2,4-Hexadienal, is used in the synthesis of chiral cycloadducts. 2,4-Hexadienal is used as a flavouring agent, as a chemical intermediate in various organic synthetic reactions and as a raw material in the manufacture of sorbic acid. It is also used as in the manufacture of polymethine dyes, as a pharmaceutical intermediate in the manufacture of mitomycins and antihypercholesteraemics, as an inhibitor of corrosion for steel used in oil field operations, as a monomer in reactions with silane comonomers for the manufacture of polyalkenyloxysilane polymer and as a fumigant against larvae of the Caribbean fruit fly.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,4-Hexadienal, is used in the synthesis of chiral cycloadducts. 2,4-Hexadienal is used as a flavouring agent, as a chemical intermediate in various organic synthetic reactions and as a raw material in the manufacture of sorbic acid. It is also used as in the manufacture of polymethine dyes, as a pharmaceutical intermediate in the manufacture of mitomycins and antihypercholesteraemics, as an inhibitor of corrosion for steel used in oil field operations, as a monomer in reactions with silane comonomers for the manufacture of polyalkenyloxysilane polymer and as a fumigant against larvae of the Caribbean fruit fly.
Solubility
It is Insoluble in water, but soluble in ethanol.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Store under inert gas.Take measures to prevent the build up of electrostatic charge. Incompatible with strong reducing agents, strong bases, strong oxidizing agents. It is Stable, Combustible.
Chemical Identifiers
142-83-6 | |
96.129 | |
BATOPAZDIZEVQF-MQQKCMAXSA-N | |
637564 | |
(2E,4E)-hexa-2,4-dienal |
C6H8O | |
MFCD00007004 | |
2,4-hexadienal, hexa-2,4-dienal, sorbaldehyde, sorbic aldehyde, trans,trans-2,4-hexadienal, 2e,4e-hexa-2,4-dienal, 2,4-hexadienal, 2e,4e, 2,4-hexadienal, e,e, 2-propyleneacrolein, 3-propyleneacrolein | |
CHEBI:82334 | |
CC=CC=CC=O |
Specifications
142-83-6 | |
170°C to 173°C | |
Floral | |
1.5386 | |
5 g | |
1698401 | |
2,4-hexadienal, hexa-2,4-dienal, sorbaldehyde, sorbic aldehyde, trans,trans-2,4-hexadienal, 2e,4e-hexa-2,4-dienal, 2,4-hexadienal, 2e,4e, 2,4-hexadienal, e,e, 2-propyleneacrolein, 3-propyleneacrolein | |
BATOPAZDIZEVQF-MQQKCMAXSA-N | |
(2E,4E)-hexa-2,4-dienal | |
637564 | |
96.13 | |
2,4-Hexadienal, predominantly trans,trans |
0.871 | |
67°C (152°F) | |
C6H8O | |
MFCD00007004 | |
UN2922 | |
Air sensitive | |
It is Insoluble in water,but soluble in ethanol. | |
CC=CC=CC=O | |
96.129 | |
CHEBI:82334 | |
95% |
Safety and Handling
GHS H Statement
H301-H311-H314-H318-H227-H317
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
May cause an allergic skin reaction.
P201-P202-P210-P264b-P270-P280i-P281-P301+P312-P302+P352-P305+P351+P338-P308+P313-P312-P330-P332+P313-P361-P363-P370+P378q-P501c
H227-H302-H311-H315-H319-H351
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 205-564-3
RTECSNumber : WG1925000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only