missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(4-Hydroxyphenylazo)benzoic acid, 97%, Thermo Scientific Chemicals

$96.56 - $456.86

Chemical Identifiers

CAS 1634-82-8
Molecular Formula C13H10N2O3
Molecular Weight (g/mol) 242.234
MDL Number MFCD00002428
InChI Key FBVSMDPNVYJNON-UHFFFAOYSA-N
Synonym 2-4-hydroxyphenylazo benzoic acid, haba, 2-p-hydroxyphenylazo benzoic acid, benzoic acid, 2-4-hydroxyphenyl azo, o-p-hydroxyphenylazo benzoic acid, 2-4'-hydroxyphenyl-azo benzoic acid, hbaba, 2-4-hydroxyphenyl diazenyl benzoic acid, 2-2-4-hydroxyphenyl diazen-1-yl benzoic acid, benzoic acid, o-p-hydroxyphenyl azo
PubChem CID 5357439
IUPAC Name 2-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]benzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)NN=C2C=CC(=O)C=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1033309
View Documents
Thermo Scientific Chemicals
A1033309
10 g
Each for $96.56
 
AAA1033318
View Documents
Thermo Scientific Chemicals
A1033318
50 g
Each for $456.86
 
Description

Description

Indicator used in determination of serum albumin

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1634-82-8
242.234
FBVSMDPNVYJNON-UHFFFAOYSA-N
5357439
C1=CC=C(C(=C1)C(=O)O)NN=C2C=CC(=O)C=C2
C13H10N2O3
MFCD00002428
2-4-hydroxyphenylazo benzoic acid, haba, 2-p-hydroxyphenylazo benzoic acid, benzoic acid, 2-4-hydroxyphenyl azo, o-p-hydroxyphenylazo benzoic acid, 2-4'-hydroxyphenyl-azo benzoic acid, hbaba, 2-4-hydroxyphenyl diazenyl benzoic acid, 2-2-4-hydroxyphenyl diazen-1-yl benzoic acid, benzoic acid, o-p-hydroxyphenyl azo
2-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]benzoic acid

Specifications

1634-82-8
C13H10N2O3
10 g
2-4-hydroxyphenylazo benzoic acid, haba, 2-p-hydroxyphenylazo benzoic acid, benzoic acid, 2-4-hydroxyphenyl azo, o-p-hydroxyphenylazo benzoic acid, 2-4'-hydroxyphenyl-azo benzoic acid, hbaba, 2-4-hydroxyphenyl diazenyl benzoic acid, 2-2-4-hydroxyphenyl diazen-1-yl benzoic acid, benzoic acid, o-p-hydroxyphenyl azo
C1=CC=C(C(=C1)C(=O)O)NN=C2C=CC(=O)C=C2
242.234
242.23
2- (4-Hydroxyphenylazo)benzoic acid
205°C to 207°C
MFCD00002428
1842696
FBVSMDPNVYJNON-UHFFFAOYSA-N
2-[2-(4-oxocyclohexa-2,5-dien-1-ylidene)hydrazinyl]benzoic acid
5357439
≥98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 216-655-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only