missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-[4-(Methoxycarbonyl)-5-methyl-2-oxo-2,3-dihydro-1H-pyrrol-3-yl]acetic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 77978-73-5
Molecular Formula C9H11NO5
Molecular Weight (g/mol) 213.189
MDL Number MFCD00173859
InChI Key WLKUVVIAHUXPSB-UHFFFAOYSA-N
Synonym 2-4-methoxycarbonyl-5-methyl-2-oxo-2,3-dihydro-1h-pyrrol-3-yl acetic acid, 4-methoxycarbonyl-5-methyl-2-oxo-1,3-dihydropyrrol-3-yl acetic acid, 1h-pyrrole-3-aceticacid, 2,3-dihydro-4-methoxycarbonyl-5-methyl-2-oxo, maybridge1_002247, 3-methoxycarbonyl-2-methyl-5-oxo-2-pyrrolin-4-yl acetic acid, 2-4-methoxycarbonyl-5-methyl-2-oxo-1,3-dihydropyrrol-3-yl acetic acid, 2-4-methoxycarbonyl-5-methyl-2-oxo-2,3-dihydro-1h-pyrrol-3yl acetic acid
PubChem CID 2774998
IUPAC Name 2-(4-methoxycarbonyl-5-methyl-2-oxo-1,3-dihydropyrrol-3-yl)acetic acid
SMILES CC1=C(C(C(=O)N1)CC(=O)O)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BTB09316DA
SDS View Documents
Thermo Scientific Chemicals
BTB09316DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB09316EA
SDS View Documents
Thermo Scientific Chemicals
BTB09316EA
10g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

77978-73-5
213.189
WLKUVVIAHUXPSB-UHFFFAOYSA-N
2774998
CC1=C(C(C(=O)N1)CC(=O)O)C(=O)OC
C9H11NO5
MFCD00173859
2-4-methoxycarbonyl-5-methyl-2-oxo-2,3-dihydro-1h-pyrrol-3-yl acetic acid, 4-methoxycarbonyl-5-methyl-2-oxo-1,3-dihydropyrrol-3-yl acetic acid, 1h-pyrrole-3-aceticacid, 2,3-dihydro-4-methoxycarbonyl-5-methyl-2-oxo, maybridge1_002247, 3-methoxycarbonyl-2-methyl-5-oxo-2-pyrrolin-4-yl acetic acid, 2-4-methoxycarbonyl-5-methyl-2-oxo-1,3-dihydropyrrol-3-yl acetic acid, 2-4-methoxycarbonyl-5-methyl-2-oxo-2,3-dihydro-1h-pyrrol-3yl acetic acid
2-(4-methoxycarbonyl-5-methyl-2-oxo-1,3-dihydropyrrol-3-yl)acetic acid

Specifications

77978-73-5
100.0
Amber Glass Bottle
MFCD00173859
2-4-methoxycarbonyl-5-methyl-2-oxo-2,3-dihydro-1h-pyrrol-3-yl acetic acid, 4-methoxycarbonyl-5-methyl-2-oxo-1,3-dihydropyrrol-3-yl acetic acid, 1h-pyrrole-3-aceticacid, 2,3-dihydro-4-methoxycarbonyl-5-methyl-2-oxo, maybridge1_002247, 3-methoxycarbonyl-2-methyl-5-oxo-2-pyrrolin-4-yl acetic acid, 2-4-methoxycarbonyl-5-methyl-2-oxo-1,3-dihydropyrrol-3-yl acetic acid, 2-4-methoxycarbonyl-5-methyl-2-oxo-2,3-dihydro-1h-pyrrol-3yl acetic acid
CC1=C(C(C(=O)N1)CC(=O)O)C(=O)OC
213.189
213.19
2-[4-(Methoxycarbonyl)-5-methyl-2-oxo-2, 3-dihydro-1H-pyrrol-3-yl]acetic acid
96.5
97%
C9H11NO5
1g
WLKUVVIAHUXPSB-UHFFFAOYSA-N
2-(4-methoxycarbonyl-5-methyl-2-oxo-1,3-dihydropyrrol-3-yl)acetic acid
2774998
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'