Learn More
2-(4-Pyridyl)thiazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals
$96.71 - $302.53
Chemical Identifiers
CAS | 21278-86-4 |
---|---|
Molecular Formula | C9H5N2O2S |
Molecular Weight (g/mol) | 205.21 |
MDL Number | MFCD00171745 |
InChI Key | COOQMBOJAAZEIR-UHFFFAOYSA-M |
Synonym | 2-pyridin-4-yl thiazole-4-carboxylic acid, 2-4-pyridyl thiazole-4-carboxylic acid, 2-pyridin-4-yl-1,3-thiazole-4-carboxylic acid, 2-4-pyridyl-1,3-thiazole-4-carboxylic acid, 2-4-pyridinyl-1,3-thiazole-4-carboxylic acid, 2-pyridin-4-yl-thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 2-4-pyridinyl, acmc-1cr1v, 4-thiazolecarboxylic acid, 2-4-pyridinyl, 4-carboxy-2-4-pyridyl thiazole |
PubChem CID | 716091 |
SMILES | [O-]C(=O)C1=CSC(=N1)C1=CC=NC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2045703
|
Thermo Scientific Chemicals
B2045703 |
1 g |
Each for $96.71
|
|
|||||
AAB2045706
|
Thermo Scientific Chemicals
B2045706 |
5 g |
Each for $302.53
|
|
|||||
Description
2-(4-Pyridyl)thiazole-4-carboxylic acid, is used as pharmaceutical intermediate and also used in the field of organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-(4-Pyridyl)thiazole-4-carboxylic acid, is used as pharmaceutical intermediate and also used in the field of organic synthesis.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal temperatures and pressures. Keep away from strong oxidizing agents, strong acids, strong bases.
Chemical Identifiers
21278-86-4 | |
205.21 | |
COOQMBOJAAZEIR-UHFFFAOYSA-M | |
716091 |
C9H5N2O2S | |
MFCD00171745 | |
2-pyridin-4-yl thiazole-4-carboxylic acid, 2-4-pyridyl thiazole-4-carboxylic acid, 2-pyridin-4-yl-1,3-thiazole-4-carboxylic acid, 2-4-pyridyl-1,3-thiazole-4-carboxylic acid, 2-4-pyridinyl-1,3-thiazole-4-carboxylic acid, 2-pyridin-4-yl-thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 2-4-pyridinyl, acmc-1cr1v, 4-thiazolecarboxylic acid, 2-4-pyridinyl, 4-carboxy-2-4-pyridyl thiazole | |
[O-]C(=O)C1=CSC(=N1)C1=CC=NC=C1 |
Specifications
21278-86-4 | |
C9H5N2O2S | |
1 g | |
Insoluble in water. | |
[O-]C(=O)C1=CSC(=N1)C1=CC=NC=C1 | |
205.21 | |
206.22 | |
2-(4-Pyridyl)thiazole-4-carboxylic acid |
>300°C (decomposition) | |
MFCD00171745 | |
2-pyridin-4-yl thiazole-4-carboxylic acid, 2-4-pyridyl thiazole-4-carboxylic acid, 2-pyridin-4-yl-1,3-thiazole-4-carboxylic acid, 2-4-pyridyl-1,3-thiazole-4-carboxylic acid, 2-4-pyridinyl-1,3-thiazole-4-carboxylic acid, 2-pyridin-4-yl-thiazole-4-carboxylic acid, 4-thiazolecarboxylicacid, 2-4-pyridinyl, acmc-1cr1v, 4-thiazolecarboxylic acid, 2-4-pyridinyl, 4-carboxy-2-4-pyridyl thiazole | |
COOQMBOJAAZEIR-UHFFFAOYSA-M | |
2-(pyridin-4-yl)-1,3-thiazole-4-carboxylate | |
716091 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only