Learn More
2,5-Bis(trifluoromethyl)benzoic acid, 98%
CAS: 42580-42-7 | C9H4F6O2 | 258.119 g/mol
Supplier: Thermo Scientific Chemicals H2618403
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 2,5-Bis(trifluoromethyl)benzoic acid | |
| 77°C to 80°C | |
| MFCD00013249 | |
| 2,5-bis trifluoromethyl benzoic acid, 2,5-di trifluoromethyl benzoic acid, 2,5-bis trifluoroformyl benzoic acid, pubchem8492, acmc-1amwk, rarechem al bo 1002, 2,5-bis-trifluoromethyl-benzoic acid, 2,5-bis trifluoromethyl benzoic acid., benzoic acid,2,5-bis trifluoromethyl, benzoic acid, 2,5-bis trifluoromethyl | |
| C1=CC(=C(C=C1C(F)(F)F)C(=O)O)C(F)(F)F | |
| 258.119 | |
| CHEBI:60697 | |
| 98% |
| 42580-42-7 | |
| C9H4F6O2 | |
| 1 g | |
| PINBPLCVZSKLTF-UHFFFAOYSA-N | |
| 2,5-bis(trifluoromethyl)benzoic acid | |
| 736187 | |
| 258.12 |
Chemical Identifiers
| 42580-42-7 | |
| 258.119 | |
| PINBPLCVZSKLTF-UHFFFAOYSA-N | |
| CHEBI:60697 | |
| C1=CC(=C(C=C1C(F)(F)F)C(=O)O)C(F)(F)F |
| C9H4F6O2 | |
| MFCD00013249 | |
| 736187 | |
| 2,5-bis(trifluoromethyl)benzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only