Learn More
Thermo Scientific Chemicals 2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol, 98%
Supplier: Thermo Scientific Chemicals 403220010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol | |
99.0 | |
155.0°C to 160.0°C | |
Authentic | |
Glass bottle | |
MFCD00006255 | |
2-5-bromo-2-pyridylazo-5-diethylamino phenol, 2-5-bromopyridin-2-yl diazenyl-5-diethylamino phenol, bromo-padap, 2-5-bromo-2-pyridylazo-5-diethylaminophenol, phenol, 2-5-bromo-2-pyridinyl azo-5-diethylamino, 2-4-diethylamino-2-hydroxyphenylazo-5-bromopyridine, hnvcxdavehoibp-vhebqxmusa-n, 2-5-bromo-2-pyridylazo-5-diethylamino-phenol, 5-diethylamino-2-5-bromo 2-pyridyl diazenyl phenol, e-2-5-bromopyridin-2-yl diazenyl-5-diethylamino phenol | |
CCN(CC)C1=CC(=O)\C(C=C1)=N\NC1=CC=C(Br)C=N1 | |
349.23 | |
349.22 | |
Crystalline Powder |
14337-53-2 | |
100.0 | |
Red | |
97.5% min. (HPLC) | |
C15H17BrN4O | |
1 g | |
LYVSIKOGJUDRBK-QGOAFFKASA-N | |
(6E)-6-[(5-bromopyridin-2-yl)hydrazinylidene]-3-(diethylamino)cyclohexa-2,4-dien-1-one | |
5911417 | |
98% |
Chemical Identifiers
14337-53-2 | |
349.23 | |
LYVSIKOGJUDRBK-QGOAFFKASA-N | |
5911417 | |
CCN(CC)C1=CC(=O)\C(C=C1)=N\NC1=CC=C(Br)C=N1 |
C15H17BrN4O | |
MFCD00006255 | |
2-5-bromo-2-pyridylazo-5-diethylamino phenol, 2-5-bromopyridin-2-yl diazenyl-5-diethylamino phenol, bromo-padap, 2-5-bromo-2-pyridylazo-5-diethylaminophenol, phenol, 2-5-bromo-2-pyridinyl azo-5-diethylamino, 2-4-diethylamino-2-hydroxyphenylazo-5-bromopyridine, hnvcxdavehoibp-vhebqxmusa-n, 2-5-bromo-2-pyridylazo-5-diethylamino-phenol, 5-diethylamino-2-5-bromo 2-pyridyl diazenyl phenol, e-2-5-bromopyridin-2-yl diazenyl-5-diethylamino phenol | |
(6E)-6-[(5-bromopyridin-2-yl)hydrazinylidene]-3-(diethylamino)cyclohexa-2,4-dien-1-one |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 238-286-6
RUO – Research Use Only