missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(5-Chloro-2-thienyl)pyrrolidine, 99%, Thermo Scientific™
$132.67 - $132.67
Chemical Identifiers
CAS | 524674-42-8 |
---|---|
Molecular Formula | C8H10ClNS |
Molecular Weight (g/mol) | 187.69 |
MDL Number | MFCD05189303 |
InChI Key | XMSYAIXTGCQRQX-UHFFFAOYNA-N |
Synonym | 2-5-chloro-2-thienyl pyrrolidine, 2-5-chlorothiophen-2-yl pyrrolidine, 2-5-chlorothien-2-yl pyrrolidine, acmc-20aoom, 2-chloro-5-pyrrolidin-2-ylthiophene, 2-5-chloro-thiophen-2-yl-pyrrolidine |
PubChem CID | 4305829 |
IUPAC Name | 2-(5-chlorothiophen-2-yl)pyrrolidine |
SMILES | ClC1=CC=C(S1)C1CCCN1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH50177MD
|
Thermo Scientific Chemicals
H50177MD |
250 mg |
Each for $132.67
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
524674-42-8 | |
187.69 | |
XMSYAIXTGCQRQX-UHFFFAOYNA-N | |
4305829 | |
ClC1=CC=C(S1)C1CCCN1 |
C8H10ClNS | |
MFCD05189303 | |
2-5-chloro-2-thienyl pyrrolidine, 2-5-chlorothiophen-2-yl pyrrolidine, 2-5-chlorothien-2-yl pyrrolidine, acmc-20aoom, 2-chloro-5-pyrrolidin-2-ylthiophene, 2-5-chloro-thiophen-2-yl-pyrrolidine | |
2-(5-chlorothiophen-2-yl)pyrrolidine |
Specifications
524674-42-8 | |
MFCD05189303 | |
2-5-chloro-2-thienyl pyrrolidine, 2-5-chlorothiophen-2-yl pyrrolidine, 2-5-chlorothien-2-yl pyrrolidine, acmc-20aoom, 2-chloro-5-pyrrolidin-2-ylthiophene, 2-5-chloro-thiophen-2-yl-pyrrolidine | |
ClC1=CC=C(S1)C1CCCN1 | |
187.69 | |
187.69 | |
2-(5-Chloro-2-thienyl)pyrrolidine |
C8H10ClNS | |
250 mg | |
XMSYAIXTGCQRQX-UHFFFAOYNA-N | |
2-(5-chlorothiophen-2-yl)pyrrolidine | |
4305829 | |
99% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures