missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-(5-Chloro-3-methylbenzo[b]thiophen-2-yl)acetic acid, 95%, Thermo Scientific™
Chemical Identifiers
CAS | 51527-19-6 |
---|---|
Molecular Formula | C11H9ClO2S |
Molecular Weight (g/mol) | 240.70 |
MDL Number | MFCD00052300 |
InChI Key | QNJIHQOPIPJYLU-UHFFFAOYSA-N |
Synonym | 2-5-chloro-3-methylbenzo b thiophen-2-yl acetic acid, tianafac, 5-chloro-3-methylbenzo b thiophene-2-acetic acid, unii-p0t3zrk3xv, p0t3zrk3xv, 2-5-chloro-3-methyl-1-benzothiophen-2-yl acetic acid, tianafacum, tianafaco, tianafac inn |
PubChem CID | 68686 |
IUPAC Name | 2-(5-chloro-3-methyl-1-benzothiophen-2-yl)acetic acid |
SMILES | CC1=C(CC(O)=O)SC2=CC=C(Cl)C=C12 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
MWP00226CB
|
Thermo Scientific Chemicals
MWP00226CB |
250mg | Amber Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
51527-19-6 | |
240.70 | |
QNJIHQOPIPJYLU-UHFFFAOYSA-N | |
68686 | |
CC1=C(CC(O)=O)SC2=CC=C(Cl)C=C12 |
C11H9ClO2S | |
MFCD00052300 | |
2-5-chloro-3-methylbenzo b thiophen-2-yl acetic acid, tianafac, 5-chloro-3-methylbenzo b thiophene-2-acetic acid, unii-p0t3zrk3xv, p0t3zrk3xv, 2-5-chloro-3-methyl-1-benzothiophen-2-yl acetic acid, tianafacum, tianafaco, tianafac inn | |
2-(5-chloro-3-methyl-1-benzothiophen-2-yl)acetic acid |
Specifications
51527-19-6 | |
95% | |
C11H9ClO2S | |
250mg | |
QNJIHQOPIPJYLU-UHFFFAOYSA-N | |
2-(5-chloro-3-methyl-1-benzothiophen-2-yl)acetic acid | |
68686 | |
95% |
95.0 | |
Amber Glass Bottle | |
MFCD00052300 | |
2-5-chloro-3-methylbenzo b thiophen-2-yl acetic acid, tianafac, 5-chloro-3-methylbenzo b thiophene-2-acetic acid, unii-p0t3zrk3xv, p0t3zrk3xv, 2-5-chloro-3-methyl-1-benzothiophen-2-yl acetic acid, tianafacum, tianafaco, tianafac inn | |
CC1=C(CC(O)=O)SC2=CC=C(Cl)C=C12 | |
240.70 | |
240.71 | |
2-(5-Chloro-3-methylbenzo[b]thiophen-2-yl)acetic acid |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.