missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dibromo-1,1-dimethyl-3,4-diphenylsilole 98.0+%, TCI America™

$517.44 - $1544.44

Chemical Identifiers

CAS 686290-22-2
Molecular Formula C18H16Br2Si
Molecular Weight (g/mol) 420.22
MDL Number MFCD28384141
InChI Key XLVXIPAEFXBVBF-UHFFFAOYSA-N
Synonym 2,5-Dibromo-1,1-dimethyl-3,4-diphenylsilacyclopenta-2,4-diene
PubChem CID 11729466
IUPAC Name 2,5-dibromo-1,1-dimethyl-3,4-diphenyl-1H-silole
SMILES C[Si]1(C)C(Br)=C(C(=C1Br)C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D46311G
View Documents
TCI America
D46311G
1 g
Each for $517.44
 
D46315G
View Documents
TCI America
D46315G
5 g
Each for $1,544.44
 
Description

Description

Specifications

Chemical Identifiers

686290-22-2
420.22
XLVXIPAEFXBVBF-UHFFFAOYSA-N
11729466
C[Si]1(C)C(Br)=C(C(=C1Br)C1=CC=CC=C1)C1=CC=CC=C1
C18H16Br2Si
MFCD28384141
2,5-Dibromo-1,1-dimethyl-3,4-diphenylsilacyclopenta-2,4-diene
2,5-dibromo-1,1-dimethyl-3,4-diphenyl-1H-silole

Specifications

686290-22-2
Yellow
MFCD28384141
2,5-Dibromo-1,1-dimethyl-3,4-diphenylsilacyclopenta-2,4-diene
C[Si]1(C)C(Br)=C(C(=C1Br)C1=CC=CC=C1)C1=CC=CC=C1
420.22
420.22
Crystalline Powder
163°C
C18H16Br2Si
1 g
XLVXIPAEFXBVBF-UHFFFAOYSA-N
2,5-dibromo-1,1-dimethyl-3,4-diphenyl-1H-silole
11729466
≥98.0% (GC)
2,5-Dibromo-1,1-dimethyl-3,4-diphenylsilole
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
missing translation for 'documents'

missing translation for 'documents'