missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dichloro-p-xylene, 98%, Thermo Scientific Chemicals

$129.48 - $319.58

Chemical Identifiers

CAS 1124-05-6
Molecular Formula C8H8Cl2
Molecular Weight (g/mol) 175.052
MDL Number MFCD00000610
InChI Key UTGSRNVBAFCOEU-UHFFFAOYSA-N
Synonym 2,5-dichloro-p-xylene, 2,5-dichloro-1,4-dimethylbenzene, benzene, 1,4-dichloro-2,5-dimethyl, p-xylene, 2,5-dichloro, unii-7p8m86zob2, 2,5-dichloro-para-xylene, p-xylene,5-dichloro, p-xylene,2,5-dichloro, acmc-2099eq, dsstox_cid_24780
PubChem CID 70755
IUPAC Name 1,4-dichloro-2,5-dimethylbenzene
SMILES CC1=CC(=C(C=C1Cl)C)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2134414
View Documents
Thermo Scientific Chemicals
B2134414
25 g N/A
 
AAB2134422
View Documents
Thermo Scientific Chemicals
B2134422
100 g N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1124-05-6
175.052
UTGSRNVBAFCOEU-UHFFFAOYSA-N
70755
CC1=CC(=C(C=C1Cl)C)Cl
C8H8Cl2
MFCD00000610
2,5-dichloro-p-xylene, 2,5-dichloro-1,4-dimethylbenzene, benzene, 1,4-dichloro-2,5-dimethyl, p-xylene, 2,5-dichloro, unii-7p8m86zob2, 2,5-dichloro-para-xylene, p-xylene,5-dichloro, p-xylene,2,5-dichloro, acmc-2099eq, dsstox_cid_24780
1,4-dichloro-2,5-dimethylbenzene

Specifications

68°C to 70°C
221°C to 223°C
C8H8Cl2
2,5-dichloro-p-xylene, 2,5-dichloro-1,4-dimethylbenzene, benzene, 1,4-dichloro-2,5-dimethyl, p-xylene, 2,5-dichloro, unii-7p8m86zob2, 2,5-dichloro-para-xylene, p-xylene,5-dichloro, p-xylene,2,5-dichloro, acmc-2099eq, dsstox_cid_24780
CC1=CC(=C(C=C1Cl)C)Cl
175.052
175.06
2,5-Dichloro-p-xylene
1124-05-6
25 g
MFCD00000610
UTGSRNVBAFCOEU-UHFFFAOYSA-N
1,4-dichloro-2,5-dimethylbenzene
70755
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 214-387-0

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only