missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2',5'-Dichloroacetophenone, 96%, Thermo Scientific™

Chemical Identifiers

CAS 2476-37-1
Molecular Formula C8H6Cl2O
Molecular Weight (g/mol) 189.04
MDL Number MFCD00000607
InChI Key CYNFEPKQDJHIMV-UHFFFAOYSA-N
Synonym 2',5'-dichloroacetophenone, 1-2,5-dichlorophenyl ethanone, 2,5-dichloroacetophenone, ethanone, 1-2,5-dichlorophenyl, acetophenone, 2',5'-dichloro, 1-2,5-dichlorophenyl ethan-1-one, 1-acetyl-2,5-dichlorobenzene, 2,5-dichloracetophenon, pubchem10586, acmc-1cmfm
PubChem CID 75587
IUPAC Name 1-(2,5-dichlorophenyl)ethanone
SMILES CC(=O)C1=C(C=CC(=C1)Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC179420100
SDS View Documents
Thermo Scientific Chemicals
179420100
10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

2476-37-1
189.04
CYNFEPKQDJHIMV-UHFFFAOYSA-N
75587
CC(=O)C1=C(C=CC(=C1)Cl)Cl
C8H6Cl2O
MFCD00000607
2',5'-dichloroacetophenone, 1-2,5-dichlorophenyl ethanone, 2,5-dichloroacetophenone, ethanone, 1-2,5-dichlorophenyl, acetophenone, 2',5'-dichloro, 1-2,5-dichlorophenyl ethan-1-one, 1-acetyl-2,5-dichlorobenzene, 2,5-dichloracetophenon, pubchem10586, acmc-1cmfm
1-(2,5-dichlorophenyl)ethanone

Specifications

2476-37-1
100.0
118.0°C (12.0 mmHg)
Authentic
Glass bottle
1.5604 to 1.5624
MFCD00000607
1.312
CYNFEPKQDJHIMV-UHFFFAOYSA-N
1-(2,5-dichlorophenyl)ethanone
75587
96%
95.0
1.3120g/mL
>110°C
95% min. (GC)
C8H6Cl2O
Cl2C6H3COCH3
10g
2',5'-dichloroacetophenone, 1-2,5-dichlorophenyl ethanone, 2,5-dichloroacetophenone, ethanone, 1-2,5-dichlorophenyl, acetophenone, 2',5'-dichloro, 1-2,5-dichlorophenyl ethan-1-one, 1-acetyl-2,5-dichlorobenzene, 2,5-dichloracetophenon, pubchem10586, acmc-1cmfm
CC(=O)C1=C(C=CC(=C1)Cl)Cl
189.04
189.04
2', 5'-Dichloroacetophenone, 96%
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 219-605-8

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only