missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dichloroaniline, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1115136

 View more versions of this product

Catalog No. AAA1115136


Explore available promotions

Description

Description

2,5-Dichloroaniline was used in a study to develop a fast and sensitive quantitative method for the detection of some aniline derivatives by solid-phase microextraction in gas chromatography-mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dichloroaniline was used in a study to develop a fast and sensitive quantitative method for the detection of some aniline derivatives by solid-phase microextraction in gas chromatography-mass spectrometry.

Solubility
Soluble in dilute hydrochloric acid.

Notes
Air & Light Sensitive, keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

2,5-Dichloroaniline
48°C to 52°C
250°C to 251°C
C6H5Cl2N
500 g
1447438
1-amino-2,5-dichlorobenzene, azoic diazo component 3, scarlet 2g base, fast scarlet gg base, 2,5-dichloranilin, benzenamine, 2,5-dichloro, azobase dca, scarlet base gg, fast scarlet 2g, scarlet base ngg
AVYGCQXNNJPXSS-UHFFFAOYSA-N
2,5-dichloroaniline
7262
162.02
95-82-9
1.54
139°C (282°F)
MFCD00007667
UN3442
Air and light sensitive
Soluble in dilute hydrochloric acid.
NC1=CC(Cl)=CC=C1Cl
162.01
CHEBI:34245
99%

Chemical Identifiers

95-82-9
162.01
AVYGCQXNNJPXSS-UHFFFAOYSA-N
7262
2,5-dichloroaniline
C6H5Cl2N
MFCD00007667
1-amino-2,5-dichlorobenzene, azoic diazo component 3, scarlet 2g base, fast scarlet gg base, 2,5-dichloranilin, benzenamine, 2,5-dichloro, azobase dca, scarlet base gg, fast scarlet 2g, scarlet base ngg
CHEBI:34245
NC1=CC(Cl)=CC=C1Cl
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H373
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P314-P330-P361-P363-P501c

H301+H311+H331-H373-H500

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: DICHLOROANILINES, SOLID

EINECSNumber : 202-455-2

RTECSNumber : BX2610000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only