missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,5-Dimethoxytoluene 98.0+%, TCI America™
Supplier: TCI America D255225G
Specifications
2,5-Dimethoxytoluene | |
Yellow | |
C9H12O2 | |
25 g | |
IQISOVKPFBLQIQ-UHFFFAOYSA-N | |
1,4-dimethoxy-2-methylbenzene | |
90552 | |
≥98.0% (GC) |
24599-58-4 | |
218°C | |
MFCD00041901 | |
2,5-dimethoxytoluene, benzene, 1,4-dimethoxy-2-methyl, toluene, 2,5-dimethoxy, methyl hydroquinone dimethyl ester, 2-methylhydroquinone dimethyl ether, 1,4-dimethoxy-2-methyl-benzene, pubchem4543, 2,5-dimethoxy toluene, acmc-209gdc | |
CC1=C(C=CC(=C1)OC)OC | |
152.193 | |
152.19 | |
Liquid |
Chemical Identifiers
24599-58-4 | |
152.193 | |
IQISOVKPFBLQIQ-UHFFFAOYSA-N | |
90552 | |
CC1=C(C=CC(=C1)OC)OC |
C9H12O2 | |
MFCD00041901 | |
2,5-dimethoxytoluene, benzene, 1,4-dimethoxy-2-methyl, toluene, 2,5-dimethoxy, methyl hydroquinone dimethyl ester, 2-methylhydroquinone dimethyl ether, 1,4-dimethoxy-2-methyl-benzene, pubchem4543, 2,5-dimethoxy toluene, acmc-209gdc | |
1,4-dimethoxy-2-methylbenzene |
Safety and Handling
TSCA : Yes