missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dimethyl-1,3-oxazole-4-carbonyl chloride, 97%, Thermo Scientific™

Chemical Identifiers

CAS 197719-27-0
Molecular Formula C6H6ClNO2
Molecular Weight (g/mol) 159.57
MDL Number MFCD04972642
InChI Key XZXCVPKMCPXVFF-UHFFFAOYSA-N
Synonym 2,5-dimethyloxazole-4-carbonyl chloride, 4-oxazolecarbonylchloride, 2,5-dimethyl, dimethyl-1,3-oxazole-4-carbonyl chloride, 2,5-dimethyl-4-chlorocarbonyl-1,3-oxazole, 2,5-dimethyl-4-oxazolecarbonyl chloride, 4-oxazolecarbonylchloride,2,5-dimethyl, 2,5-dimethyl-oxazole-4-carbonyl chloride
PubChem CID 7018020
IUPAC Name 2,5-dimethyl-1,3-oxazole-4-carbonyl chloride
SMILES CC1=NC(C(Cl)=O)=C(C)O1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC26802CB
SDS View Documents
Thermo Scientific Chemicals
CC26802CB
250mg Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC26802DA
SDS View Documents
Thermo Scientific Chemicals
CC26802DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC26802DE
SDS View Documents
Thermo Scientific Chemicals
CC26802DE
5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

197719-27-0
159.57
XZXCVPKMCPXVFF-UHFFFAOYSA-N
7018020
CC1=NC(C(Cl)=O)=C(C)O1
C6H6ClNO2
MFCD04972642
2,5-dimethyloxazole-4-carbonyl chloride, 4-oxazolecarbonylchloride, 2,5-dimethyl, dimethyl-1,3-oxazole-4-carbonyl chloride, 2,5-dimethyl-4-chlorocarbonyl-1,3-oxazole, 2,5-dimethyl-4-oxazolecarbonyl chloride, 4-oxazolecarbonylchloride,2,5-dimethyl, 2,5-dimethyl-oxazole-4-carbonyl chloride
2,5-dimethyl-1,3-oxazole-4-carbonyl chloride

Specifications

197719-27-0
100.0
Amber Glass Bottle
MFCD04972642
2,5-dimethyloxazole-4-carbonyl chloride, 4-oxazolecarbonylchloride, 2,5-dimethyl, dimethyl-1,3-oxazole-4-carbonyl chloride, 2,5-dimethyl-4-chlorocarbonyl-1,3-oxazole, 2,5-dimethyl-4-oxazolecarbonyl chloride, 4-oxazolecarbonylchloride,2,5-dimethyl, 2,5-dimethyl-oxazole-4-carbonyl chloride
CC1=NC(C(Cl)=O)=C(C)O1
159.57
159.57
2, 5-Dimethyl-1, 3-oxazole-4-carbonyl chloride
96.5
97%
C6H6ClNO2
250mg
XZXCVPKMCPXVFF-UHFFFAOYSA-N
2,5-dimethyl-1,3-oxazole-4-carbonyl chloride
7018020
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/eye protection/face protection.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
missing translation for 'documents'

missing translation for 'documents'