missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,5-Dimethyl-1,4-phenylenediamine 98.0+%, TCI America™
$177.75 - $515.48
Chemical Identifiers
CAS | 6393-01-7 |
---|---|
Molecular Formula | C8H12N2 |
Molecular Weight (g/mol) | 136.20 |
MDL Number | MFCD00052675 |
InChI Key | BWAPJIHJXDYDPW-UHFFFAOYSA-N |
Synonym | 2,5-dimethyl-1,4-benzenediamine, 4-amino-2,5-dimethylaniline, 2,5-dimethyl-1,4-phenylenediamine, 2,5-dimethyl-p-phenylenediamine, 2,5-diamino-p-xylene, 2,5-dimethylphenylene-1,4-diamine, 2,5-dimethyl-1,4-benzoquinonediamine, ccris 8118, 1,4-dimethylphenylene-2,5-diamine, unii-re400vn2b4 |
PubChem CID | 160824 |
ChEBI | CHEBI:53613 |
IUPAC Name | 2,5-dimethylbenzene-1,4-diamine |
SMILES | CC1=CC(=C(C=C1N)C)N |
Chemical Identifiers
6393-01-7 | |
136.20 | |
BWAPJIHJXDYDPW-UHFFFAOYSA-N | |
160824 | |
2,5-dimethylbenzene-1,4-diamine |
C8H12N2 | |
MFCD00052675 | |
2,5-dimethyl-1,4-benzenediamine, 4-amino-2,5-dimethylaniline, 2,5-dimethyl-1,4-phenylenediamine, 2,5-dimethyl-p-phenylenediamine, 2,5-diamino-p-xylene, 2,5-dimethylphenylene-1,4-diamine, 2,5-dimethyl-1,4-benzoquinonediamine, ccris 8118, 1,4-dimethylphenylene-2,5-diamine, unii-re400vn2b4 | |
CHEBI:53613 | |
CC1=CC(=C(C=C1N)C)N |
Specifications
148°C | |
25 g | |
C8H12N2 | |
2,5-dimethyl-1,4-benzenediamine, 4-amino-2,5-dimethylaniline, 2,5-dimethyl-1,4-phenylenediamine, 2,5-dimethyl-p-phenylenediamine, 2,5-diamino-p-xylene, 2,5-dimethylphenylene-1,4-diamine, 2,5-dimethyl-1,4-benzoquinonediamine, ccris 8118, 1,4-dimethylphenylene-2,5-diamine, unii-re400vn2b4 | |
CC1=CC(=C(C=C1N)C)N | |
136.20 | |
CHEBI:53613 | |
≥98.0% (GC,T) |
6393-01-7 | |
Crystal-Powder at 20°C | |
MFCD00052675 | |
BWAPJIHJXDYDPW-UHFFFAOYSA-N | |
2,5-dimethylbenzene-1,4-diamine | |
160824 | |
136.20 | |
2,5-Dimethyl-1,4-phenylenediamine |
Safety and Handling
TSCA : No