missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dimethyl-2,5-di(tert-butylperoxy)hexane, 92%, Thermo Scientific Chemicals

$66.55 - $131.48

Chemical Identifiers

CAS 78-63-7
Molecular Formula C16H34O4
Molecular Weight (g/mol) 290.44
InChI Key DMWVYCCGCQPJEA-UHFFFAOYSA-N
Synonym varox, 2,5-dimethyl-2,5-di tert-butylperoxy hexane, 2,5-bis tert-butylperoxy-2,5-dimethylhexane, triqanox xq 8, kayahexa ad 40c, lupersol 101, luperco 101xl, luperox 101, perhexa 3m40, varox 50
PubChem CID 6545
IUPAC Name 2,5-bis(tert-butylperoxy)-2,5-dimethylhexane
SMILES CC(C)(C)OOC(C)(C)CCC(C)(C)OOC(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC349900050
SDS View Documents
Thermo Scientific Chemicals
349900050
5 g Plastic bottle
Each for $66.55
 
AC349901000
SDS View Documents
Thermo Scientific Chemicals
349901000
100 g Plastic bottle
Each for $131.48
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Radical initiators
Specifications

Chemical Identifiers

78-63-7
290.44
varox, 2,5-dimethyl-2,5-di tert-butylperoxy hexane, 2,5-bis tert-butylperoxy-2,5-dimethylhexane, triqanox xq 8, kayahexa ad 40c, lupersol 101, luperco 101xl, luperox 101, perhexa 3m40, varox 50
2,5-bis(tert-butylperoxy)-2,5-dimethylhexane
C16H34O4
DMWVYCCGCQPJEA-UHFFFAOYSA-N
6545
CC(C)(C)OOC(C)(C)CCC(C)(C)OOC(C)(C)C

Specifications

78-63-7
0.8700g/mL
68°C
Plastic bottle
1.4200 to 1.4240
0.87
Solubility in water: immiscible. Other solubilities: soluble in organic solvents,soluble in alcohol
CC(C)(C)OOC(C)(C)CCC(C)(C)OOC(C)(C)C
290.44
6.4 mPa.s (20°C)
92%
6.0°C
55.0°C to 57.0°C (7.0 mmHg)
90% min. (GC)
C16H34O4
5 g
varox, 2,5-dimethyl-2,5-di tert-butylperoxy hexane, 2,5-bis tert-butylperoxy-2,5-dimethylhexane, triqanox xq 8, kayahexa ad 40c, lupersol 101, luperco 101xl, luperox 101, perhexa 3m40, varox 50
DMWVYCCGCQPJEA-UHFFFAOYSA-N
2,5-bis(tert-butylperoxy)-2,5-dimethylhexane
6545
290.44
2, 5-Dimethyl-2, 5-di(tert-butylperoxy)hexane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Heating may cause a fire.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep/Store away from clothing/ combustible materials.
Store in a well-v

GHS Signal Word: Danger

EINECSNumber : 201-128-1

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only