missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,5-Dimethyl-2,5-hexanediol, 97%, Thermo Scientific Chemicals
$99.95 - $99.95
Chemical Identifiers
CAS | 110-03-2 |
---|---|
Molecular Formula | C8H18O2 |
Molecular Weight (g/mol) | 146.23 |
MDL Number | MFCD00004473 |
InChI Key | ZWNMRZQYWRLGMM-UHFFFAOYSA-N |
Synonym | 2,5-dimethyl-2,5-hexanediol, 2,5-hexanediol, 2,5-dimethyl, unii-3pnb2s8721, 1,1,4,4-tetramethyl-1,4-butanediol, 2,5-dihydroxy-2,5-dimethylhexane, 2,5-dimethyl-hexane-2,5-diol, dimethylhexanediol, dsstox_cid_6830, dsstox_rid_78222, dsstox_gsid_26830 |
PubChem CID | 8031 |
IUPAC Name | 2,5-dimethylhexane-2,5-diol |
SMILES | CC(C)(O)CCC(C)(C)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC162502500
|
Thermo Scientific Chemicals
162502500 |
250 g | Glass bottle |
Each for $99.95
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
110-03-2 | |
146.23 | |
ZWNMRZQYWRLGMM-UHFFFAOYSA-N | |
8031 | |
CC(C)(O)CCC(C)(C)O |
C8H18O2 | |
MFCD00004473 | |
2,5-dimethyl-2,5-hexanediol, 2,5-hexanediol, 2,5-dimethyl, unii-3pnb2s8721, 1,1,4,4-tetramethyl-1,4-butanediol, 2,5-dihydroxy-2,5-dimethylhexane, 2,5-dimethyl-hexane-2,5-diol, dimethylhexanediol, dsstox_cid_6830, dsstox_rid_78222, dsstox_gsid_26830 | |
2,5-dimethylhexane-2,5-diol |
Specifications
110-03-2 | |
GC≥96.0% | |
88°C to 91°C | |
214°C to 215°C | |
Conforms | |
Glass bottle | |
HOC(CH3)2CH2CH2C(CH3)2OH | |
250 g | |
2,5-dimethyl-2,5-hexanediol, 2,5-hexanediol, 2,5-dimethyl, unii-3pnb2s8721, 1,1,4,4-tetramethyl-1,4-butanediol, 2,5-dihydroxy-2,5-dimethylhexane, 2,5-dimethyl-hexane-2,5-diol, dimethylhexanediol, dsstox_cid_6830, dsstox_rid_78222, dsstox_gsid_26830 | |
ZWNMRZQYWRLGMM-UHFFFAOYSA-N | |
2,5-dimethylhexane-2,5-diol | |
8031 | |
97% | |
2, 5-Dimethyl-2, 5-hexanediol, 97% |
98.5 | |
100.0 | |
White | |
126°C | |
96% min. (GC) | |
C8H18O2 | |
MFCD00004473 | |
01, 492 | |
Solubility in water: soluble. Other solubilities: soluble in acetone,ethylene glycol,insoluble in benzene,ccl4,petroleuw ether | |
CC(C)(O)CCC(C)(C)O | |
146.23 | |
146.23 | |
Flakes, Pellets or Prills |
Safety and Handling
GHS H Statement
Causes serious eye damage.
GHS Signal Word: Danger
EINECSNumber : 203-731-5
TSCA : TSCA
RUO – Research Use Only