missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethyl-3-furanthiol 97.0+%, TCI America™
$104.38 - $313.16
Chemical Identifiers
| CAS | 55764-23-3 |
|---|---|
| Molecular Formula | C6H8OS |
| Molecular Weight (g/mol) | 128.19 |
| MDL Number | MFCD00801026 |
| InChI Key | DBBHCZMXKBCICL-UHFFFAOYSA-N |
| Synonym | 2,5-dimethyl-3-furanthiol, 3-mercapto-2,5-dimethylfuran, 3-furanthiol, 2,5-dimethyl, unii-wct2259x5l, 2,5-dimethyl-3-mercaptofuran, fema no. 3451, 2,5-dimethyl-3-furylmercaptan, 2,5-dimethyl-3-furan thiol, 2,5-dimethyl-furan-3-thiol, acmc-1amqa |
| PubChem CID | 41569 |
| IUPAC Name | 2,5-dimethylfuran-3-thiol |
| SMILES | CC1=CC(S)=C(C)O1 |
Chemical Identifiers
| 55764-23-3 | |
| 128.19 | |
| DBBHCZMXKBCICL-UHFFFAOYSA-N | |
| 41569 | |
| CC1=CC(S)=C(C)O1 |
| C6H8OS | |
| MFCD00801026 | |
| 2,5-dimethyl-3-furanthiol, 3-mercapto-2,5-dimethylfuran, 3-furanthiol, 2,5-dimethyl, unii-wct2259x5l, 2,5-dimethyl-3-mercaptofuran, fema no. 3451, 2,5-dimethyl-3-furylmercaptan, 2,5-dimethyl-3-furan thiol, 2,5-dimethyl-furan-3-thiol, acmc-1amqa | |
| 2,5-dimethylfuran-3-thiol |
Specifications
| 55764-23-3 | |
| C6H8OS | |
| 1 g | |
| 2,5-dimethyl-3-furanthiol, 3-mercapto-2,5-dimethylfuran, 3-furanthiol, 2,5-dimethyl, unii-wct2259x5l, 2,5-dimethyl-3-mercaptofuran, fema no. 3451, 2,5-dimethyl-3-furylmercaptan, 2,5-dimethyl-3-furan thiol, 2,5-dimethyl-furan-3-thiol, acmc-1amqa | |
| CC1=CC(S)=C(C)O1 | |
| 128.19 | |
| 128.19 | |
| Liquid |
| 150°C | |
| MFCD00801026 | |
| 3336 | |
| DBBHCZMXKBCICL-UHFFFAOYSA-N | |
| 2,5-dimethylfuran-3-thiol | |
| 41569 | |
| ≥97.0% (GC) | |
| 2,5-Dimethyl-3-furanthiol |
Safety and Handling
RTECSNumber : LU6230000
TSCA : No