Learn More
2,5-Dimethyl-3-hexyne-2,5-diol, (+/-) + meso, 98%, Thermo Scientific Chemicals
$80.38 - $233.55
Chemical Identifiers
CAS | 142-30-3 |
---|---|
Molecular Formula | C8H14O2 |
Molecular Weight (g/mol) | 142.198 |
MDL Number | MFCD00004468 |
InChI Key | IHJUECRFYCQBMW-UHFFFAOYSA-N |
Synonym | 2,5-dimethyl-3-hexyne-2,5-diol, dimethylhexynediol, kemitracin-50, tetramethylbutynediol, acetylenepinacol, 3-hexyne-2,5-diol, 2,5-dimethyl, olfine y, tetramethyl-2-butynediol, unii-22rr53u71w, 2,5-dimethylhexyne-2,5-diol |
PubChem CID | 8883 |
IUPAC Name | 2,5-dimethylhex-3-yne-2,5-diol |
SMILES | CC(C)(C#CC(C)(C)O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2088922
|
Thermo Scientific Chemicals
B2088922 |
100 g |
Each for $80.38
|
|
|||||
AAB2088936
|
Thermo Scientific Chemicals
B2088936 |
500 g |
Each for $233.55
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
142-30-3 | |
142.198 | |
IHJUECRFYCQBMW-UHFFFAOYSA-N | |
8883 | |
CC(C)(C#CC(C)(C)O)O |
C8H14O2 | |
MFCD00004468 | |
2,5-dimethyl-3-hexyne-2,5-diol, dimethylhexynediol, kemitracin-50, tetramethylbutynediol, acetylenepinacol, 3-hexyne-2,5-diol, 2,5-dimethyl, olfine y, tetramethyl-2-butynediol, unii-22rr53u71w, 2,5-dimethylhexyne-2,5-diol | |
2,5-dimethylhex-3-yne-2,5-diol |
Specifications
142-30-3 | |
0.949 | |
102°C (215°F) | |
MFCD00004468 | |
969835 | |
IHJUECRFYCQBMW-UHFFFAOYSA-N | |
2,5-dimethylhex-3-yne-2,5-diol | |
8883 | |
98% |
90°C to 94°C | |
205°C | |
C8H14O2 | |
100 g | |
2,5-dimethyl-3-hexyne-2,5-diol, dimethylhexynediol, kemitracin-50, tetramethylbutynediol, acetylenepinacol, 3-hexyne-2,5-diol, 2,5-dimethyl, olfine y, tetramethyl-2-butynediol, unii-22rr53u71w, 2,5-dimethylhexyne-2,5-diol | |
CC(C)(C#CC(C)(C)O)O | |
142.198 | |
142.2 | |
2,5-Dimethyl-3-hexyne-2,5-diol, (+/-) + meso |
Safety and Handling
GHS H Statement
H317
May cause an allergic skin reaction.
GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
EINECSNumber : 205-533-4
RTECSNumber : MR0176000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only