missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dimethylbenzaldehyde, 98%, Thermo Scientific Chemicals

$438.29 - $438.29

Chemical Identifiers

CAS 5779-94-2
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00003342
InChI Key SMUVABOERCFKRW-UHFFFAOYSA-N
PubChem CID 22015
IUPAC Name 2,5-dimethylbenzaldehyde
SMILES CC1=CC=C(C)C(C=O)=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0192414
View Documents
Thermo Scientific Chemicals
L0192414
25 g
Each for $438.29
 
Description

Description

2,5-Dimethylbenzaldehyde is useful for building asymmetric top molecules. 2,5-DMBA has also been used to analyze vehicle carbon emissions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dimethylbenzaldehyde is useful for building asymmetric top molecules. 2,5-DMBA has also been used to analyze vehicle carbon emissions.

Solubility
Not miscible in water.

Notes
Air Sensitive. Store under dry inert gas. Do not expose to air. Incompatible with Air, strong bases, strong oxidizing agents.
Specifications

Chemical Identifiers

5779-94-2
134.18
SMUVABOERCFKRW-UHFFFAOYSA-N
2,5-dimethylbenzaldehyde
C9H10O
MFCD00003342
22015
CC1=CC=C(C)C(C=O)=C1

Specifications

5779-94-2
104°C to 106°C (14 mmHg)
Almond-like
1.544
25 g
Air Sensitive
Not miscible in water.
CC1=CC=C(C)C(C=O)=C1
134.18
134.18
2,5-Dimethylbenzaldehyde
0.95
87°C (188°F)
C9H10O
MFCD00003342
1927303
benzaldehyde, 2,5-dimethyl, isoxylaldehyde, 2,5-dimethyl-benzaldehyde, acmc-20aoaf, pubchem1474, benzaldehyde,2,5-dimethyl, 2,5-dimethylbenzaldehyde
SMUVABOERCFKRW-UHFFFAOYSA-N
2,5-dimethylbenzaldehyde
22015
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P280-P370+P378q-P501c

H227

EINECSNumber : 227-303-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only