missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Dimethylterephthalic Acid 97.0+%, TCI America™

$610.27 - $2583.30

Chemical Identifiers

CAS 6051-66-7
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00020292
InChI Key FKUJGZJNDUGCFU-UHFFFAOYSA-N
PubChem CID 299989
IUPAC Name 2,5-dimethylterephthalic acid
SMILES CC1=CC(=C(C=C1C(=O)O)C)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D22085G
View Documents
TCI America
D22085G
5 g
Each for $610.27
 
D220825G
View Documents
TCI America
D220825G
25 g
Each for $2,583.30
 
Description

Description

Specifications

Chemical Identifiers

6051-66-7
194.186
FKUJGZJNDUGCFU-UHFFFAOYSA-N
2,5-dimethylterephthalic acid
C10H10O4
MFCD00020292
299989
CC1=CC(=C(C=C1C(=O)O)C)C(=O)O

Specifications

6051-66-7
C10H10O4
5 g
CC1=CC(=C(C=C1C(=O)O)C)C(=O)O
194.186
194.19
Crystalline Powder
White
MFCD00020292
FKUJGZJNDUGCFU-UHFFFAOYSA-N
2,5-dimethylterephthalic acid
299989
≥97.0% (GC,T)
2,5-Dimethylterephthalic Acid
Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'