missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Diphenyl-p-benzoquinone, 96%, Thermo Scientific Chemicals

$82.55 - $294.01

Chemical Identifiers

CAS 844-51-9
Molecular Formula C18H12O2
Molecular Weight (g/mol) 260.29
MDL Number MFCD00001600
InChI Key QYXHDJJYVDLECA-UHFFFAOYSA-N
Synonym 2,5-diphenyl-1,4-benzoquinone, 2,5-diphenyl-p-benzoquinone, 2,5-diphenylquinone, 2,5-diphenyl-4-benzoquinone, 2,5-diphenylbenzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-diphenyl, 1,1':4',1-terphenyl-2',5'-dione, p-benzoquinone, 2,5-diphenyl, acmc-209pup, 2,5-diphenyl-p-quinone
PubChem CID 70055
IUPAC Name 2,5-diphenylcyclohexa-2,5-diene-1,4-dione
SMILES O=C1C=C(C(=O)C=C1C1=CC=CC=C1)C1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1536906
View Documents
Thermo Scientific Chemicals
A1536906
5 g
Each for $82.55
 
AAA1536914
View Documents
Thermo Scientific Chemicals
A1536914
25 g
Each for $294.01
 
Description

Description

2,5-Diphenyl-p-benzoquinone is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Diphenyl-p-benzoquinone is used as pharmaceutical intermediate.

Solubility
Soluble in hot Toluene (almost transparency).

Notes
Incompatible with Oxidizing agents, Reducing agent.
Specifications

Chemical Identifiers

844-51-9
260.29
QYXHDJJYVDLECA-UHFFFAOYSA-N
70055
O=C1C=C(C(=O)C=C1C1=CC=CC=C1)C1=CC=CC=C1
C18H12O2
MFCD00001600
2,5-diphenyl-1,4-benzoquinone, 2,5-diphenyl-p-benzoquinone, 2,5-diphenylquinone, 2,5-diphenyl-4-benzoquinone, 2,5-diphenylbenzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-diphenyl, 1,1':4',1-terphenyl-2',5'-dione, p-benzoquinone, 2,5-diphenyl, acmc-209pup, 2,5-diphenyl-p-quinone
2,5-diphenylcyclohexa-2,5-diene-1,4-dione

Specifications

844-51-9
C18H12O2
5 g
2,5-diphenyl-1,4-benzoquinone, 2,5-diphenyl-p-benzoquinone, 2,5-diphenylquinone, 2,5-diphenyl-4-benzoquinone, 2,5-diphenylbenzoquinone, 2,5-cyclohexadiene-1,4-dione, 2,5-diphenyl, 1,1':4',1-terphenyl-2',5'-dione, p-benzoquinone, 2,5-diphenyl, acmc-209pup, 2,5-diphenyl-p-quinone
QYXHDJJYVDLECA-UHFFFAOYSA-N
2,5-diphenylcyclohexa-2,5-diene-1,4-dione
70055
96%
215°C to 219°C
MFCD00001600
2051812
Soluble in hot Toluene (almost transparency).
O=C1C=C(C(=O)C=C1C1=CC=CC=C1)C1=CC=CC=C1
260.29
260.3
2,5-Diphenyl-p-benzoquinone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c

H315-H317-H319-H334-H335

EINECSNumber : 212-680-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only