missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,5-Hexanedione, 97%, Thermo Scientific Chemicals

$71.22 - $420.03

Chemical Identifiers

CAS 110-13-4
Molecular Formula C6H10O2
Molecular Weight (g/mol) 114.144
MDL Number MFCD00008792
InChI Key OJVAMHKKJGICOG-UHFFFAOYSA-N
Synonym 2,5-hexanedione, acetonylacetone, 1,2-diacetylethane, acetonyl acetone, diacetonyl, 2,5-diketohexane, 2,5-hexadione, acetone, acetonyl, 2,5-hexandione, alpha,beta-diacetylethane
PubChem CID 8035
ChEBI CHEBI:85014
IUPAC Name hexane-2,5-dione
SMILES CC(=O)CCC(=O)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2568614
View Documents
Thermo Scientific Chemicals
B2568614
25 g
Each for $71.22
 
AAB2568622
View Documents
Thermo Scientific Chemicals
B2568622
100 g
Each for $134.22
 
AAB2568636
View Documents
Thermo Scientific Chemicals
B2568636
500 g
Each for $420.03
 
Description

Description

2,5-Hexanedione is used as a reagent in the preparation of trans-2,5-dimethylpyrrolidine. It is also used in the synthesis of 2,5-dimethylpyrroles. Further, it plays an important role as a reagent used for the protection of amino groups in amino sugars and nucleosides. In addition to this, it is used in the preparation of five-membered heterocycles like indane-type and benzannulated systems. It is also employed as a precursor in Diels-alder cycloaddition reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Hexanedione is used as a reagent in the preparation of trans-2,5-dimethylpyrrolidine. It is also used in the synthesis of 2,5-dimethylpyrroles. Further, it plays an important role as a reagent used for the protection of amino groups in amino sugars and nucleosides. In addition to this, it is used in the preparation of five-membered heterocycles like indane-type and benzannulated systems. It is also employed as a precursor in Diels-alder cycloaddition reactions.

Solubility
Miscible with alcohol, water and diethyl ether.

Notes
Incompatible with strong oxidizing agents, strong bases, metals and strong reducing agents.
Specifications

Chemical Identifiers

110-13-4
114.144
OJVAMHKKJGICOG-UHFFFAOYSA-N
8035
hexane-2,5-dione
C6H10O2
MFCD00008792
2,5-hexanedione, acetonylacetone, 1,2-diacetylethane, acetonyl acetone, diacetonyl, 2,5-diketohexane, 2,5-hexadione, acetone, acetonyl, 2,5-hexandione, alpha,beta-diacetylethane
CHEBI:85014
CC(=O)CCC(=O)C

Specifications

110-13-4
0.976
78°C (172°F)
1.425
25 g
14,71
Miscible with alcohol,water and diethyl ether.
CC(=O)CCC(=O)C
114.144
CHEBI:85014
97%
-6°C
192°C to 193°C
C6H10O2
MFCD00008792
506525
2,5-hexanedione, acetonylacetone, 1,2-diacetylethane, acetonyl acetone, diacetonyl, 2,5-diketohexane, 2,5-hexadione, acetone, acetonyl, 2,5-hexandione, alpha,beta-diacetylethane
OJVAMHKKJGICOG-UHFFFAOYSA-N
hexane-2,5-dione
8035
114.15
2,5-Hexanedione
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H361-H373-H302-H332-H315-H319-H335-H227
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

P210-P260-P264b-P280-P302+P352-P305+P351+P338-P314-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H373

EINECSNumber : 203-738-3

RTECSNumber : MO3150000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only