missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
5-Methyltryptamine, hydrochloride, 98%, Thermo Scientific™
$234.07 - $234.07
Chemical Identifiers
Molecular Formula | C11H15N2 |
---|---|
Molecular Weight (g/mol) | 175.25 |
MDL Number | MFCD00012683 |
InChI Key | PYOUAIQXJALPKW-UHFFFAOYSA-O |
Synonym | 5-methyltryptamine hydrochloride, 2-5-methyl-1h-indol-3-yl ethanamine hydrochloride, 3-2-aminoethyl-5-methylindole hydrochloride, 5-methyl-1h-indole-3-ethylamine monohydrochloride, 5-methyltryptamine hcl, 1h-indole-3-ethanamine, 5-methyl-, monohydrochloride, 2-5-methylindol-3-yl ethylamine, chloride, 5-methyltryptaminehydrochloride, 5-methyl-1h-indole-3-ethylamine hcl, pubchem24287 |
PubChem CID | 6917145 |
SMILES | CC1=CC=C2NC=C(CC[NH3+])C2=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC158670010
|
Thermo Scientific Chemicals
158670010 |
1 g |
Each for $234.07
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
C11H15N2 | |
MFCD00012683 | |
5-methyltryptamine hydrochloride, 2-5-methyl-1h-indol-3-yl ethanamine hydrochloride, 3-2-aminoethyl-5-methylindole hydrochloride, 5-methyl-1h-indole-3-ethylamine monohydrochloride, 5-methyltryptamine hcl, 1h-indole-3-ethanamine, 5-methyl-, monohydrochloride, 2-5-methylindol-3-yl ethylamine, chloride, 5-methyltryptaminehydrochloride, 5-methyl-1h-indole-3-ethylamine hcl, pubchem24287 | |
CC1=CC=C2NC=C(CC[NH3+])C2=C1 |
175.25 | |
PYOUAIQXJALPKW-UHFFFAOYSA-O | |
6917145 |
Specifications
1010-95-3 | |
100.0 | |
Beige to Brown | |
97.5% min. (Argentometry) | |
C11H15N2 | |
1 g | |
PYOUAIQXJALPKW-UHFFFAOYSA-O | |
2-(5-methyl-1H-indol-3-yl)ethanamine;hydrochloride | |
6917145 | |
98% | |
5-Methyltryptamine,hydrochloride |
97.5 | |
281°C to 283°C | |
Authentic | |
Glass bottle | |
MFCD00012683 | |
5-methyltryptamine hydrochloride, 2-5-methyl-1h-indol-3-yl ethanamine hydrochloride, 3-2-aminoethyl-5-methylindole hydrochloride, 5-methyl-1h-indole-3-ethylamine monohydrochloride, 5-methyltryptamine hcl, 1h-indole-3-ethanamine, 5-methyl-, monohydrochloride, 2-5-methylindol-3-yl ethylamine, chloride, 5-methyltryptaminehydrochloride, 5-methyl-1h-indole-3-ethylamine hcl, pubchem24287 | |
CC1=CC=C2NC=C(CC[NH3+])C2=C1 | |
175.25 | |
210.71 | |
Crystalline Powder |
Safety and Handling
EINECSNumber : 213-777-8
RUO – Research Use Only