missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(5-Methyl-2-phenyl-1,3-oxazol-4-yl)acetic acid, 97%, Thermo Scientific™

Chemical Identifiers

CAS 107367-98-6
Molecular Formula C12H11NO3
Molecular Weight (g/mol) 217.224
MDL Number MFCD00100005
InChI Key XEWJNPORMBGGKZ-UHFFFAOYSA-N
Synonym 2-5-methyl-2-phenyl-1,3-oxazol-4-yl acetic acid, 5-methyl-2-phenyl-1,3-oxazol-4-yl acetic acid, 2-5-methyl-2-phenyloxazol-4-yl acetic acid, 5-methyl-2-phenyloxazol-4-yl acetic acid, maybridge1_002118, acmc-2098vv, 2-phenyl-5-methyl-4-oxazolyiacetic acid, 2-phenyl-5-methyl-4-oxazolylacetic acid, 2-phenyl-5-methyloxazol-4-ylacetic acid, 5-methyl-2-phenyl-1,3-oxazolyl acetic acid
PubChem CID 2775139
IUPAC Name 2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)acetic acid
SMILES CC1=C(N=C(O1)C2=CC=CC=C2)CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
BTB09929DA
SDS View Documents
Thermo Scientific Chemicals
BTB09929DA
1g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
BTB09929EB
SDS View Documents
Thermo Scientific Chemicals
BTB09929EB
25g N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

107367-98-6
217.224
XEWJNPORMBGGKZ-UHFFFAOYSA-N
2775139
CC1=C(N=C(O1)C2=CC=CC=C2)CC(=O)O
C12H11NO3
MFCD00100005
2-5-methyl-2-phenyl-1,3-oxazol-4-yl acetic acid, 5-methyl-2-phenyl-1,3-oxazol-4-yl acetic acid, 2-5-methyl-2-phenyloxazol-4-yl acetic acid, 5-methyl-2-phenyloxazol-4-yl acetic acid, maybridge1_002118, acmc-2098vv, 2-phenyl-5-methyl-4-oxazolyiacetic acid, 2-phenyl-5-methyl-4-oxazolylacetic acid, 2-phenyl-5-methyloxazol-4-ylacetic acid, 5-methyl-2-phenyl-1,3-oxazolyl acetic acid
2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)acetic acid

Specifications

107367-98-6
100.0
Amber Glass Bottle
MFCD00100005
2-5-methyl-2-phenyl-1,3-oxazol-4-yl acetic acid, 5-methyl-2-phenyl-1,3-oxazol-4-yl acetic acid, 2-5-methyl-2-phenyloxazol-4-yl acetic acid, 5-methyl-2-phenyloxazol-4-yl acetic acid, maybridge1_002118, acmc-2098vv, 2-phenyl-5-methyl-4-oxazolyiacetic acid, 2-phenyl-5-methyl-4-oxazolylacetic acid, 2-phenyl-5-methyloxazol-4-ylacetic acid, 5-methyl-2-phenyl-1,3-oxazolyl acetic acid
CC1=C(N=C(O1)C2=CC=CC=C2)CC(=O)O
217.224
217.22
2-(5-Methyl-2-phenyl-1, 3-oxazol-4-yl)acetic acid
96.5
97%
C12H11NO3
1g
XEWJNPORMBGGKZ-UHFFFAOYSA-N
2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)acetic acid
2775139
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
missing translation for 'documents'

missing translation for 'documents'