missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(5-Methyl-2-phenyl-1,3-oxazol-4-yl)ethan-1-ol, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals BTB09930DA

 View more versions of this product

Catalog No. BTB09930DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

Specifications

Specifications

2-(5-Methyl-2-phenyl-1, 3-oxazol-4-yl)ethan-1-ol
96.5
97%
C12H13NO2
1g
JYWHQBLLIBQGCU-UHFFFAOYSA-N
2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)ethanol
725585
97%
103788-65-4
100.0
Amber glass bottle
MFCD00100006
2-5-methyl-2-phenyl-1,3-oxazol-4-yl ethan-1-ol, 2-5-methyl-2-phenyloxazol-4-yl ethanol, 2-5-methyl-2-phenyl-1,3-oxazol-4-yl ethanol, 4-2-hydroxyethyl-5-methyl-2-phenyloxazol, 4-oxazoleethanol, 5-methyl-2-phenyl, 2-5-methyl-2-phenyl-oxazol-4-yl-ethanol, 4-2-hydroxyethyl-5-methyl-2-phenyl-1,3-oxazole, 4-oxazoleethanol,5-methyl-2-phenyl, 2-5-methyl-2-phenyl-oxazol-4-yl ethanol, 2-5-methyl-2-phenyl-1,3-oxazol-4-yl-1-ethanol
CC1=C(N=C(O1)C2=CC=CC=C2)CCO
203.241
203.24

Chemical Identifiers

103788-65-4
203.241
JYWHQBLLIBQGCU-UHFFFAOYSA-N
725585
CC1=C(N=C(O1)C2=CC=CC=C2)CCO
C12H13NO2
MFCD00100006
2-5-methyl-2-phenyl-1,3-oxazol-4-yl ethan-1-ol, 2-5-methyl-2-phenyloxazol-4-yl ethanol, 2-5-methyl-2-phenyl-1,3-oxazol-4-yl ethanol, 4-2-hydroxyethyl-5-methyl-2-phenyloxazol, 4-oxazoleethanol, 5-methyl-2-phenyl, 2-5-methyl-2-phenyl-oxazol-4-yl-ethanol, 4-2-hydroxyethyl-5-methyl-2-phenyl-1,3-oxazole, 4-oxazoleethanol,5-methyl-2-phenyl, 2-5-methyl-2-phenyl-oxazol-4-yl ethanol, 2-5-methyl-2-phenyl-1,3-oxazol-4-yl-1-ethanol
2-(5-methyl-2-phenyl-1,3-oxazol-4-yl)ethanol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS Documents

Documents

Promotions

Promotions