missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Di-tert-butylpyridine 97.0+%, TCI America™
$264.70 - $911.52
Chemical Identifiers
| CAS | 585-48-8 |
|---|---|
| Molecular Formula | C13H21N |
| Molecular Weight (g/mol) | 191.318 |
| MDL Number | MFCD00006306 |
| InChI Key | UWKQJZCTQGMHKD-UHFFFAOYSA-N |
| Synonym | 2,6-di-tert-butylpyridine, 2,6-di tert-butyl pyridine, 2,6-di-t-butylpyridine, unii-oi9lf0h4mm, 2,6-di-t-butyl pyridine, pyridine, 2,6-bis 1,1-dimethylethyl, oi9lf0h4mm, 2,6-di-tert-butyl pyridine, pyridine, 2,6-di-tert-butyl, pubchem16119 |
| PubChem CID | 68510 |
| IUPAC Name | 2,6-ditert-butylpyridine |
| SMILES | CC(C)(C)C1=NC(=CC=C1)C(C)(C)C |
Chemical Identifiers
| 585-48-8 | |
| 191.318 | |
| UWKQJZCTQGMHKD-UHFFFAOYSA-N | |
| 68510 | |
| CC(C)(C)C1=NC(=CC=C1)C(C)(C)C |
| C13H21N | |
| MFCD00006306 | |
| 2,6-di-tert-butylpyridine, 2,6-di tert-butyl pyridine, 2,6-di-t-butylpyridine, unii-oi9lf0h4mm, 2,6-di-t-butyl pyridine, pyridine, 2,6-bis 1,1-dimethylethyl, oi9lf0h4mm, 2,6-di-tert-butyl pyridine, pyridine, 2,6-di-tert-butyl, pubchem16119 | |
| 2,6-ditert-butylpyridine |
Specifications
| 585-48-8 | |
| Yellow | |
| C13H21N | |
| 5 g | |
| UWKQJZCTQGMHKD-UHFFFAOYSA-N | |
| 2,6-ditert-butylpyridine | |
| 68510 | |
| ≥97.0% (GC) | |
| 2,6-Di-tert-butylpyridine |
| 2°C | |
| 101°C | |
| MFCD00006306 | |
| 2,6-di-tert-butylpyridine, 2,6-di tert-butyl pyridine, 2,6-di-t-butylpyridine, unii-oi9lf0h4mm, 2,6-di-t-butyl pyridine, pyridine, 2,6-bis 1,1-dimethylethyl, oi9lf0h4mm, 2,6-di-tert-butyl pyridine, pyridine, 2,6-di-tert-butyl, pubchem16119 | |
| CC(C)(C)C1=NC(=CC=C1)C(C)(C)C | |
| 191.318 | |
| 191.32 | |
| Liquid |
Safety and Handling
TSCA : No