missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2,6-Diaminopimelic acid, 96%, Thermo Scientific Chemicals
$120.58 - $309.38
Chemical Identifiers
CAS | 583-93-7 |
---|---|
Molecular Formula | C7H14N2O4 |
Molecular Weight (g/mol) | 190.199 |
MDL Number | MFCD00002637 |
InChI Key | GMKMEZVLHJARHF-UHFFFAOYSA-N |
Synonym | 2,6-diaminopimelic acid, heptanedioic acid, 2,6-diamino, diaminopimelic acid, dl-2,6-diaminoheptanedioic acid, m-dap, dl-alpha,epsilon-diaminopimelic acid, 1,5-diaminoheptanedioic acid, ll-diaminopimelate, d,l-diaminopimelate, ll-a2pm |
PubChem CID | 865 |
ChEBI | CHEBI:23673 |
IUPAC Name | 2,6-diaminoheptanedioic acid |
SMILES | C(CC(C(=O)O)N)CC(C(=O)O)N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAB2239103
|
Thermo Scientific Chemicals
B2239103 |
1 g |
Each for $120.58
|
|
|||||
AAB2239106
|
Thermo Scientific Chemicals
B2239106 |
5 g |
Each for $309.38
|
|
|||||
Description
2,6-Diaminopimelic acid is used as a constituent of proteoglycans. It is used as a component of bacterial cell wall.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-Diaminopimelic acid is used as a constituent of proteoglycans. It is used as a component of bacterial cell wall.
Solubility
Soluble in water, diluted acids and alkali.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
583-93-7 | |
190.199 | |
GMKMEZVLHJARHF-UHFFFAOYSA-N | |
865 | |
2,6-diaminoheptanedioic acid |
C7H14N2O4 | |
MFCD00002637 | |
2,6-diaminopimelic acid, heptanedioic acid, 2,6-diamino, diaminopimelic acid, dl-2,6-diaminoheptanedioic acid, m-dap, dl-alpha,epsilon-diaminopimelic acid, 1,5-diaminoheptanedioic acid, ll-diaminopimelate, d,l-diaminopimelate, ll-a2pm | |
CHEBI:23673 | |
C(CC(C(=O)O)N)CC(C(=O)O)N |
Specifications
583-93-7 | |
0.98 | |
MFCD00002637 | |
2,6-diaminopimelic acid, heptanedioic acid, 2,6-diamino, diaminopimelic acid, dl-2,6-diaminoheptanedioic acid, m-dap, dl-alpha,epsilon-diaminopimelic acid, 1,5-diaminoheptanedioic acid, ll-diaminopimelate, d,l-diaminopimelate, ll-a2pm | |
GMKMEZVLHJARHF-UHFFFAOYSA-N | |
2,6-diaminoheptanedioic acid | |
865 | |
190.2 |
>300°C | |
C7H14N2O4 | |
1 g | |
Soluble in water,diluted acids and alkali. | |
C(CC(C(=O)O)N)CC(C(=O)O)N | |
190.199 | |
CHEBI:23673 | |
2, 6-Diaminopimelic acid |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 209-524-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only