Learn More
2,6-Diaminopyridine, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 112385000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2, 6-Diaminopyridine | |
141-86-6 | |
100.0 | |
155°C | |
97.5% min. (GC) | |
C5H7N3 | |
MFCD00006329 | |
22,I,647 | |
Solubility in water: 9.9g/100 mL (20°C) | |
C1=CC(=NC(=C1)N)N | |
109.13 | |
109.13 |
98% | |
97.5 | |
285°C | |
Authentic | |
Glass bottle | |
(C5H3N)(NH2)2 | |
500g | |
2,6-diaminopyridine, 2,6-pyridinediamine, 2,6-diamino pyridine, pyridine, 2,6-diamino, dap amine, pyridine-2,6-diyldiamine, unii-k89ab8ckg6, ccris 6682, k89ab8ckg6, dsstox_cid_20127 | |
VHNQIURBCCNWDN-UHFFFAOYSA-N | |
pyridine-2,6-diamine | |
8861 | |
98% |
Safety and Handling
GHS H Statement
Toxic if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume
GHS Signal Word: Danger
EINECSNumber : 205-507-2
RTECSNumber : US7570000
TSCA : TSCA
Recommended Storage : Product darkens on storage
RUO â Research Use Only