Learn More
2,6-Diaminotoluene, 97%, Thermo Scientific Chemicals
$63.55 - $157.42
Chemical Identifiers
CAS | 823-40-5 |
---|---|
Molecular Formula | C7H10N2 |
Molecular Weight (g/mol) | 122.17 |
MDL Number | MFCD00007800 |
InChI Key | RLYCRLGLCUXUPO-UHFFFAOYSA-N |
Synonym | 2,6-diaminotoluene, 2,6-toluenediamine, 1,3-benzenediamine, 2-methyl, toluene-2,6-diamine, 2,6-tolylenediamine, 2,6-toluylenediamine, 2,6-diamino-1-methylbenzene, 2-methyl-1,3-benzenediamine, 2-methyl-1,3-phenylenediamine, 2-methyl-m-phenylenediamine |
PubChem CID | 13205 |
ChEBI | CHEBI:76288 |
IUPAC Name | 2-methylbenzene-1,3-diamine |
SMILES | CC1=C(N)C=CC=C1N |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0427806
|
Thermo Scientific Chemicals
L0427806 |
5 g |
Each for $63.55
|
|
|||||
AAL0427814
|
Thermo Scientific Chemicals
L0427814 |
25 g |
Each for $157.42
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
823-40-5 | |
122.17 | |
RLYCRLGLCUXUPO-UHFFFAOYSA-N | |
13205 | |
2-methylbenzene-1,3-diamine |
C7H10N2 | |
MFCD00007800 | |
2,6-diaminotoluene, 2,6-toluenediamine, 1,3-benzenediamine, 2-methyl, toluene-2,6-diamine, 2,6-tolylenediamine, 2,6-toluylenediamine, 2,6-diamino-1-methylbenzene, 2-methyl-1,3-benzenediamine, 2-methyl-1,3-phenylenediamine, 2-methyl-m-phenylenediamine | |
CHEBI:76288 | |
CC1=C(N)C=CC=C1N |
Specifications
823-40-5 | |
C7H10N2 | |
5 g | |
2079476 | |
RLYCRLGLCUXUPO-UHFFFAOYSA-N | |
2-methylbenzene-1,3-diamine | |
13205 | |
122.17 | |
2,6-Diaminotoluene |
104°C to 106°C | |
MFCD00007800 | |
UN2811 | |
2,6-diaminotoluene, 2,6-toluenediamine, 1,3-benzenediamine, 2-methyl, toluene-2,6-diamine, 2,6-tolylenediamine, 2,6-toluylenediamine, 2,6-diamino-1-methylbenzene, 2-methyl-1,3-benzenediamine, 2-methyl-1,3-phenylenediamine, 2-methyl-m-phenylenediamine | |
CC1=C(N)C=CC=C1N | |
122.17 | |
CHEBI:76288 | |
97% |
Safety and Handling
GHS H Statement
H341-H302-H312-H317
Suspected of causing genetic defects.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
P201-P202-P261-P264b-P270-P272-P280g-P281-P301+P312-P302+P352-P308+P313-P312-P330-P333+P313-P363-P501c
H302+H312-H317-H341
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 212-513-9
RTECSNumber : XS9750000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only