missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dichlorobenzoyl chloride, 99%, Thermo Scientific Chemicals

$110.78 - $110.78

Chemical Identifiers

CAS 4659-45-4
Molecular Formula C7H3Cl3O
Molecular Weight (g/mol) 209.46
MDL Number MFCD00000662
InChI Key JBLIDPPHFGWTKU-UHFFFAOYSA-N
Synonym benzoyl chloride, 2,6-dichloro, 2,6-dichlorobenzoylchloride, unii-394f6s81wy, 2,6-dichloro-benzoyl chloride, ccris 8617, pubchem18674, acmc-209k7i, 2.6-dichlorobenzoylchloride, dsstox_cid_12891, dsstox_rid_79043
PubChem CID 78392
IUPAC Name 2,6-dichlorobenzoyl chloride
SMILES C1=CC(=C(C(=C1)Cl)C(=O)Cl)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC182060100
SDS View Documents
Thermo Scientific Chemicals
182060100
10 mL Glass bottle
Each for $110.78
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

4659-45-4
209.46
JBLIDPPHFGWTKU-UHFFFAOYSA-N
78392
C1=CC(=C(C(=C1)Cl)C(=O)Cl)Cl
C7H3Cl3O
MFCD00000662
benzoyl chloride, 2,6-dichloro, 2,6-dichlorobenzoylchloride, unii-394f6s81wy, 2,6-dichloro-benzoyl chloride, ccris 8617, pubchem18674, acmc-209k7i, 2.6-dichlorobenzoylchloride, dsstox_cid_12891, dsstox_rid_79043
2,6-dichlorobenzoyl chloride

Specifications

4659-45-4
100.0
Colorless to Yellow or Pink
142°C to 143°C (21.0 mmHg)
Authentic
Glass bottle
1.5590 to 1.561
MFCD00000662
09, 343
benzoyl chloride, 2,6-dichloro, 2,6-dichlorobenzoylchloride, unii-394f6s81wy, 2,6-dichloro-benzoyl chloride, ccris 8617, pubchem18674, acmc-209k7i, 2.6-dichlorobenzoylchloride, dsstox_cid_12891, dsstox_rid_79043
JBLIDPPHFGWTKU-UHFFFAOYSA-N
2,6-dichlorobenzoyl chloride
78392
99%
2, 6-Dichlorobenzoyl chloride, 99%
98.5
18°C
1.4500g/mL
>110°C
98.5% min. (GC)
C7H3Cl3O
Cl2C6H3COCl
10 mL
1.45
Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether and acetone,slightly soluble in heptane
C1=CC(=C(C(=C1)Cl)C(=O)Cl)Cl
209.46
209.46
Liquid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 225-102-4

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only