missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2,6-Dichloroindophenol, sodium salt hydrate, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 152870250

 View more versions of this product

Catalog No. AC152870250


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2, 6-Dichloroindophenol, sodium salt hydrate
98+%
MFCD00150014
Solubility in water: soluble. Other solubilities: freely soluble in alcohol
Authentic
sodium 4-[(3,5-dichloro-4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzen-1-olate
23696612
290.07
Glass bottle
Dark Green
Powder
1266615-56-8
C12H6Cl2NNaO2
Tillman's reagent hydrate
CVSUAFOWIXUYQA-UHFFFAOYSA-M
[Na+].[O-]C1=CC=C(C=C1)N=C1C=C(Cl)C(=O)C(Cl)=C1
290.07
14% max. (1 g, 120°C)
≥98%
For determination of ascorbic acid: Passes Test
25 g

Chemical Identifiers

1266615-56-8
290.07
CVSUAFOWIXUYQA-UHFFFAOYSA-M
23696612
C12H6Cl2NNaO2
MFCD00150014
Tillman's reagent hydrate
[Na+].[O-]C1=CC=C(C=C1)N=C1C=C(Cl)C(=O)C(Cl)=C1
Videos
SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only