Learn More
2,6-Difluoro-3-nitrotoluene, 98%, Thermo Scientific™
Chemical Identifiers
CAS | 79562-49-5 |
---|---|
Molecular Formula | C7H5F2NO2 |
Molecular Weight (g/mol) | 173.119 |
MDL Number | MFCD11519030 |
InChI Key | AVWNNVJXXMKAPB-UHFFFAOYSA-N |
Synonym | 2,6-difluoro-3-nitrotoluene, benzene, 1,3-difluoro-2-methyl-4-nitro, 2,4-difluoro-3-methylnitrobenzene, pubchem10410, pubchem15439, 1,3-difluoro-2-methyl-4-nitro-benzene, 1,3-bis fluoranyl-2-methyl-4-nitro-benzene |
PubChem CID | 12795486 |
IUPAC Name | 1,3-difluoro-2-methyl-4-nitrobenzene |
SMILES | CC1=C(C=CC(=C1F)[N+](=O)[O-])F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH63431MD
|
Thermo Scientific Chemicals
H63431MD |
250 mg |
N/A
|
|
|||||
AAH6343103
|
Thermo Scientific Chemicals
H6343103 |
1 g |
N/A
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications2,6-Difluoro-3-nitrotoluene used as a intermediate in organic synthesis, pharmaceutical and chemical research.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Chemical Identifiers
79562-49-5 | |
173.119 | |
AVWNNVJXXMKAPB-UHFFFAOYSA-N | |
12795486 | |
CC1=C(C=CC(=C1F)[N+](=O)[O-])F |
C7H5F2NO2 | |
MFCD11519030 | |
2,6-difluoro-3-nitrotoluene, benzene, 1,3-difluoro-2-methyl-4-nitro, 2,4-difluoro-3-methylnitrobenzene, pubchem10410, pubchem15439, 1,3-difluoro-2-methyl-4-nitro-benzene, 1,3-bis fluoranyl-2-methyl-4-nitro-benzene | |
1,3-difluoro-2-methyl-4-nitrobenzene |
Specifications
79562-49-5 | |
MFCD11519030 | |
UN2811 | |
Slightly soluble in water. | |
CC1=C(C=CC(=C1F)[N+](=O)[O-])F | |
173.119 | |
173.12 | |
Solid |
C7H5F2NO2 | |
250 mg | |
2,6-difluoro-3-nitrotoluene, benzene, 1,3-difluoro-2-methyl-4-nitro, 2,4-difluoro-3-methylnitrobenzene, pubchem10410, pubchem15439, 1,3-difluoro-2-methyl-4-nitro-benzene, 1,3-bis fluoranyl-2-methyl-4-nitro-benzene | |
AVWNNVJXXMKAPB-UHFFFAOYSA-N | |
1,3-difluoro-2-methyl-4-nitrobenzene | |
12795486 | |
98% | |
2,6-Difluoro-3-nitrotoluene |
Safety and Handling
GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures